Skip to Content
Merck
All Photos(1)

Key Documents

F3665

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O111:B4

FITC conjugate

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

Escherichia coli (O111:B4)

Quality Level

conjugate

FITC conjugate

form

lyophilized powder

extent of labeling

2-10 μg FITC per mg LPS

color

yellow to orange

solubility

H2O: soluble 4.90-5.10 mg/mL, faintly hazy to hazy, pale yellow to yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

FITC-Lipopolysaccharides (FITC-LPS) may be used to study processes such as the movement/transport of LPS across tissue barriers such as the colonic epithelium and cellular binding and internalization of lipopolysaccharides with fluorescence-based assay systems. It may be used to evaluate, qualify and quantify lipopolysaccharides (LPS) removal reagents and procedures. FITC-LPS conjugate may be used for the detection of LPA binding proteins and motifs.[1][2]
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) yielding a hazy, yellow to orange solution. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution. For cell culture use, LPS should be reconstituted by adding 1 ml of sterile balanced salt solution or cell culture medium to a vial (1 mg) and swirling gently until the powder dissolves. Solutions can be further diluted to the desired working concentration with additional sterile balanced salt solutions or cell culture media.

Other Notes

To gain a comprehensive understanding of our extensive range of lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yannick Boege et al.
Cancer cell, 32(3), 342-359 (2017-09-13)
Concomitant hepatocyte apoptosis and regeneration is a hallmark of chronic liver diseases (CLDs) predisposing to hepatocellular carcinoma (HCC). Here, we mechanistically link caspase-8-dependent apoptosis to HCC development via proliferation- and replication-associated DNA damage. Proliferation-associated replication stress, DNA damage, and genetic
Saurabh Srivastava et al.
Antimicrobial agents and chemotherapy, 57(6), 2457-2466 (2013-03-13)
Temporin L (TempL) is a 13-residue frog antimicrobial peptide that shows moderate bactericidal activity and antiendotoxin properties in macrophages. We envisioned that, due to its very hydrophobic nature, the peptide might fail to show its desired biological properties. It was
Bin Li et al.
International journal of molecular sciences, 15(1), 1143-1161 (2014-01-21)
Innate immunity is the first line of defense in human beings against pathogen infection; monocytes/macrophages are the primary cells of the innate immune system. Recently, macrophages/monocytes have been discovered to participate in LPS clearance, and the clearance efficiency determines the
Amit Kumar et al.
Scientific reports, 7, 39925-39925 (2017-01-05)
To become clinically effective, antimicrobial peptides (AMPs) should be non-cytotoxic to host cells. Piscidins are a group of fish-derived AMPs with potent antimicrobial and antiendotoxin activities but limited by extreme cytotoxicity. We conjectured that introduction of cationic residue(s) at the
Li Liu et al.
Journal of cellular and molecular medicine, 19(12), 2728-2740 (2015-08-21)
It remains unclear whether and how cardiomyocytes contribute to the inflammation in chronic heart failure (CHF). We recently reviewed the capacity of cardiomyocytes to initiate inflammation, by means of expressing certain immune receptors such as toll-like receptors (TLRs) that respond

Articles

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service