Skip to Content
Merck
All Photos(1)

Key Documents

F2883

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–FITC antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Mouse IgG FITC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

sheep

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:128

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Mouse IgG (whole molecule) F(ab′)2 fragment-FITC antibody is specific for ) F(ab′)2 fragment of mouse IgG1, G2a, G2b, and G3 subclasses. The antibody preparation is solid phase adsorbed with human serum proteins prior to conjugation to minimize cross reactivity. The antibody preparation is conjugated to Fluorescein Isothiocyanate (FITC), Isomer I.
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment is specific for mouse IgG subclasses G1, G2a, G2b, and G3 as demonstrated by Ouchterlony double diffusion using mouse myeloma proteins. The antibody product can be used to avoid background staining due to the presence of Fc receptors.

Specificity

Binds all mouse Igs.
Useful when trying to avoid background staining due to the presence of Fc receptors.

Immunogen

Mouse IgG
Pepsin digest of antiserum

Application

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment-FITC antibody may be used in direct immunofluorescence at a minimum antibody dilution of 1:128. The antibody was used at a working dilution of 1:25 to stain microtubules in epidermal cells of Vigna unguiculata or cowpea It was also used for flow cytometry analysis using COS cells and human peripheral blood mononuclear cells.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuliana Napolitano et al.
Mutation research, 749(1-2), 21-27 (2013-08-03)
Double strand DNA breaks (DSBs) are one of the most challenging forms of DNA damage which, if left unrepaired, can trigger cellular death and can contribute to cancer. A number of studies have been focused on DNA-damage response (DDR) mechanisms
D Skalamera et al.
The Plant journal : for cell and molecular biology, 16(2), 191-200 (1998-10-01)
During the infection of cowpea (Vigna unguiculata) by the cowpea rust fungus (Uromyces vignae, race 1 ) the plant nucleus moves towards and away from the invading hypha and eventually moves close to the fungus in the susceptible cultivar while
J R Huth et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(10), 5231-5236 (2000-05-11)
The leukocyte integrin, lymphocyte function-associated antigen 1 (LFA-1) (CD11a/CD18), mediates cell adhesion and signaling in inflammatory and immune responses. To support these functions, LFA-1 must convert from a resting to an activated state that avidly binds its ligands such as
Andrea Lunardi et al.
Developmental biology, 290(2), 411-420 (2006-01-13)
Dystroglycan (DG) is a transmembrane receptor linking the extracellular matrix to the internal cytoskeleton. Its structural function has been mainly characterized in muscle fibers, but DG plays signaling and developmental roles also in different tissues and cell types. We have
Min Zhao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 16(8), 857-859 (2002-04-23)
Directed cell migration is essential for tissue formation, inflammation, and wound healing. Chemotaxis plays a major role in these situations and is underpinned by asymmetric intracellular signaling. Endogenous electric fields (EFs) are common where cell movement occurs, such as in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service