Skip to Content
Merck
All Photos(1)

Key Documents

F0791

Sigma-Aldrich

Anti-Fibronectin Antibody

mouse monoclonal, IST-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-Fibronectin antibody produced in mouse, clone IST-3, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

IST-3, monoclonal

mol wt

antigen 240 kDa

contains

15 mM sodium azide

species reactivity

goat, canine, human, chicken, sheep, rabbit, bovine

technique(s)

immunocytochemistry: suitable
immunohistochemistry (frozen sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: 1:50 using cultured human fibroblasts
microarray: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Fibronectin (FN) is an extracellular matrix protein composed of two nearly-identical disulfide-bound polypeptides with typical molecular weights of 220-280 kDa. Cellular fibronectin is structurally and antigenically similar to cold insoluble globulin from plasma and antibodies to either form usually cross-react. Careful analysis of the FN molecule indicates that it contains several functionally and structurally distinct domains which may bind to cell surfaces and to a variety of molecules such as collagen, heparin, gelatin, fibrin and DNA. FN′s play an important role in diverse biological phenomena including cell adhesion, cell migration, hemostasis and thrombosis, wound healing and the ability to induce a more normal phenotype in transformed cells.
Monoclonal Anti-Human Fibronectin (mouse IgG1 iso-type) is derived from the IST-3 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c immunized mice. Fibronectin (FN) is a multifunctional extracellular matrix protein comprising two nearly-identical disulfide-bound polypeptides. It is an adhesive glycoprotein. FN belongs to the ligand glycoprotein family and is is expressed in various cell types. Human fibronectin consists of five distinct domains that are referred to as heparin (Hep)-1/ fibrin (Fib)-1, gelatin (Gel), cell surface (Cell), Hep-2 and Fib-2, based on their affinity for Hep, Gel, Cell or Fib. The FN gene is mapped to human chromosome location 2q35.

Specificity

Monoclonal Anti-Human Fibronectin, clone IST-3, recognizes an epitope located within the fourth type-three repeat of human plasma fibronectin.

Immunogen

human plasma fibronectin

Application

Mouse monoclonal clone IST-3 anti-Human Fibronectin antibody has been used in:
  • enzyme linked immuno sorbent assay (ELISA)
  • western blotting
  • dot blot
  • radio immunoassay (RIA)
  • immunocytochemistry
  • immunohistochemistry
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Fibronectin (FN) binds to cell surfaces and a variety of molecules, such as collagen, heparin, gelatin, fibrin, and DNA. It plays an important role in diverse biological phenomena including cell adhesion, cell migration, hemostasis, and thrombosis. FN is also involved in wound healing and stimulates a normal phenotype in transformed cells. Upregulation of FN leads to tumors, such as nasopharyngeal carcinoma (NPC), ovarian cancer, and head and neck squamous cell carcinoma.

Physical form

The product is provided as ascites fluid containing 0.1% sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Zardi et al.
European journal of biochemistry, 146(3), 571-579 (1985-02-01)
Human plasma fibronectin is composed of at least five distinct domains which we refer to as Hep-1/Fib-1, Gel, Cell, Hep-2 and Fib-2 depending on their affinity for heparin (Hep), gelatin (Gel), the cell surface (Cell) or fibrin (Fib). These domains
Cellular distribution of invadopodia is regulated by nanometer scale surface
protein patterns
Ayaz GB, et al.
European Journal of Cell Biology, 673-684 (2017)
Mariana Izquierdo et al.
BioMed research international, 2014, 781246-781246 (2014-09-02)
Adherence to the intestinal epithelia is a key feature in enteroaggregative Escherichia coli (EAEC) infection. The aggregative adherence fimbriae (AAFs) are involved in EAEC interaction with receptors at the surface of intestinal cells. We and others have demonstrated that fibronectin
Asuka Nanbo et al.
The Journal of general virology, 97(11), 2989-3006 (2016-09-23)
Epstein-Barr virus (EBV) establishes a lifelong latent infection in B lymphocytes and often is found in epithelial cells. Several lines of evidence indicate that viral transmission mediated by cell-to-cell contact is the dominant mode of infection by EBV for epithelial
Sahel N Abduljauwad et al.
Scientific reports, 9(1), 5935-5935 (2019-04-13)
Cancer metastasis results from the suppression of adhesion between cancer cells and the extracellular matrix, causing their migration from the primary tumor location and the subsequent formation of tumors in distant organs. This study demonstrates the potential use of nano-sized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service