Key Documents
EPACSB-500S
Platform Clamps, for Eppendorf® Shakers
for Eppendorf® New Brunswick Innova 2000 shaker, for 500 mL media bottles
About This Item
UNSPSC Code:
41103816
NACRES:
NB.73
feature
for 500 mL media bottles
for Eppendorf® New Brunswick Innova 2000 shaker
manufacturer/tradename
Eppendorf® ACSB-500S (Global)
Related Categories
General description
Flask clamps are of a single piece stainless-steel construction (not spot welded or riveted) for added strength and safety. Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds. Flask clamps come complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG