EP951020346
Eppendorf twin.tec® PCR Plates 96
semi skirted, size 250 μL, green, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
size:
250 μL
96 well
96 well
sterility:
non-sterile
feature:
skirt
semi skirted
semi skirted
Recommended Products
sterility
non-sterile
feature
skirt
semi skirted
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 951020346
technique(s)
PCR: suitable
size
250 μL
96 well
color
green
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
Eppendorf “twin.tec” PCR Plates 96 are semi skirted PCR plates with 96 wells. The plates are green in color and are compatible with most thermal cyclers.
Application
Eppendorf “twin.tec” PCR Plates 96 supports a wide variety of PCR applications like:
- Standard PCR
- Real time PCR
- High throughput PCR
PCR
Features and Benefits
- Eppendorf [TM=“twin.tec”] PCR Plates 96 consist of wells with a maximum capacity of 250 µL.
- The wells are semi skirted and are available with barcodes.
- These plates have an enhanced well to well tolerance.
- Eppendorf [TM=“twin.tec”] PCR Plates 96 are suitable for quantitative real-time PCR.
- An [TM=“OptiTrack”] matrix supports rapid sample identification and reduces pipetting errors.
- Maximum recovery rates for improved assay results
- Available with barcode (upon request)
- Ideal for quantitative real-time PCR as well
- Exceptionally solid and torque-resistant polycarbonate frame
- OptiTrack® matrix for faster sample identification and fewer pipetting errors
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service