EP4430000018
Eppendorf® Easypet® 3 Electronic Pipette Controller
includes power supply, wallholder, removable shelf rack and 2 membrane filters 0.45 um, non-sterile
Synonym(s):
Flow indicator
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
sterility
non-sterile
feature
includes power supply, wallholder, removable shelf rack and 2 membrane filters 0.45 um
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 4430000018
Related Categories
General description
Experience a new dimension of electronic pipetting with the Easypet® 3. The latest technological developments give you complete speed control with the utmost precision. The lightweight, well-balanced Easypet 3 has been designed considering all ergonomic findings to fit comfortably in your hand and matches its shape to allow for fatigue-free pipetting.
Application
- Liquid handling with serological and volumetric pipettes from 0.1 – 100 mL.
- Serial dispensing of aliquots of different volumes.
- Resuspension of bacteria or cell pellets.
- Aspiration of cell layer from e.g. Ficoll® gradient.
Features and Benefits
- Eppendorf® Easypet® 3 electronic pipet controller is ideal for use with serological and volumetric pipettes.
- Convenient speed adjustment enables sensitive control of the meniscus.
- Ergonomically well-designed, lightweight controller for increased precision and ease of use.
- Vibrant backlit LEDs facilitate battery status display.
- Lithium polymer rechargeable battery offers long runtime without the need for recharging.
- Autoclavable pipette adapter for sterile applications.
- Quick-release aspirating cone for easy exchange of membrane filters.
- A powerful motor ensures efficient performance to speed up large volume pipetting.
Legal Information
Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Ficoll is a registered trademark of Cytiva
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service