Documents
EP0030129636
Eppendorf twin.tec® Real-Time PCR Plates 96
Skirted, Forensic DNA Grade, working volume 150 μL, white plate, pack of 10 (individually wrapped)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
material
white plate
white wells
well size
96
sterility
non-sterile
feature
skirt
Forensic DNA Grade
Skirted
packaging
pack of 10 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030129636 (Global)
technique(s)
PCR: suitable
size
96 well
working volume
150 μL
Features and Benefits
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG