Skip to Content
Merck
All Photos(1)

Key Documents

EHU152001

Sigma-Aldrich

MISSION® esiRNA

targeting human MMP14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAGAAGTTTTACGGCTTGCAAGTAACAGGCAAAGCTGATGCAGACACCATGAAGGCCATGAGGCGCCCCCGATGTGGTGTTCCAGACAAGTTTGGGGCTGAGATCAAGGCCAATGTTCGAAGGAAGCGCTACGCCATCCAGGGTCTCAAATGGCAACATAATGAAATCACTTTCTGCATCCAGAATTACACCCCCAAGGTGGGCGAGTATGCCACATACGAGGCCATTCGCAAGGCGTTCCGCGTGTGGGAGAGTGCCACACCACTGCGCTTCCGCGAGGTGCCCTATGCCTACATCCGTGAGGGCCATGAGAAGCAGGCCGACATCATGATCTTCTTTGCCGAGGGCTTCCATGGCGACAGCACGCCCTTCGATGGTGAGGGCGGCTTCCTGGCCCATGCCTACTTCCCAGGCCCCAACATTGGAGGAGACACCCACT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Po-Hsiang Chang et al.
Scientific reports, 8, 45751-45751 (2017-04-04)
Cancer stem cells (CSCs), a small population of cancer cells, have been considered to be the origin of cancer initiation, recurrence, and metastasis. Tumor microenvironment provides crucial signals for CSCs to maintain stem cell properties and promotes tumorigenesis. Therefore, establishment
Pirita Pekkonen et al.
eLife, 7 (2018-05-02)
Lymphatic invasion and lymph node metastasis correlate with poor clinical outcome in melanoma. However, the mechanisms of lymphatic dissemination in distant metastasis remain incompletely understood. We show here that exposure of expansively growing human WM852 melanoma cells, but not singly
Charles Marusak et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(22), 6039-6050 (2020-08-21)
The extracellular matrix (ECM) is an intriguing, yet understudied component of therapy resistance. Here, we investigated the role of ECM remodeling by the collagenase, MT1-MMP, in conferring resistance of v-Raf murine sarcoma viral oncogene homolog B1 (BRAF)-mutant melanoma to BRAF
Evelyne Tassone et al.
Journal of cellular physiology, 230(2), 366-377 (2014-07-06)
Membrane-type 1 matrix metalloproteinase (MT1-MMP, MMP-14), a transmembrane proteinase with an extracellular catalytic domain and a short cytoplasmic tail, degrades extracellular matrix components and controls diverse cell functions through proteolytic and non-proteolytic interactions with extracellular, intracellular, and transmembrane proteins. Here
Shawn P Carey et al.
Scientific reports, 7, 42088-42088 (2017-02-12)
A critical step in breast cancer progression is local tissue invasion, during which cells pass from the epithelial compartment to the stromal compartment. We recently showed that malignant leader cells can promote the invasion of otherwise non-invasive epithelial follower cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service