Skip to Content
Merck
All Photos(1)

Key Documents

EHU147971

Sigma-Aldrich

MISSION® esiRNA

targeting human MIF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGTGGTGTCCGAGAAGTCAGGCACGTAGCTCAGCGGCGGCCGCGGCGCGTGCGTCTGTGCCTCTGCGCGGGTCTCCTGGTCCTTCTGCCATCATGCCGATGTTCATCGTAAACACCAACGTGCCCCGCGCCTCCGTGCCGGACGGGTTCCTCTCCGAGCTCACCCAGCAGCTGGCGCAGGCCACCGGCAAGCCCCCCCAGTACATCGCGGTGCACGTGGTCCCGGACCAGCTCATGGCCTTCGGCGGCTCCAGCGAGCCGTGCGCGCTCTGCAGCCTGCACAGCATCGGCAAGATCGGCGGCGCGCAGAACCGCTCCTACAGCAAGCTGCTGTGCGGCCTGCTGGCCGAGCGCCTGCGCATCAGCCCGGACAGGGTCTACATCAACTATTACGACATGAACGCGGCCAATGTGGGCTGGAACAACTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei Zhang et al.
Molecular carcinogenesis, 58(6), 898-912 (2019-01-23)
Macrophage migration inhibitory factor (MIF) is a prominent orchestrator during the onset and progression of cancer. Recently, MIF was detected in salivary adenoid cystic carcinoma (SACC). However, its functional effect in perineural invasion (PNI) of SACC remained unknown. To illuminate
Milica Jovanović Krivokuća et al.
Reproduction, fertility, and development (2020-12-17)
Extravillous trophoblasts are specific placental cells that invade the uterine stroma and spiral arteries modifying and adjusting them to pregnancy. Many pregnancy pathologies are associated with impairment of this process, including preeclampsia and intrauterine growth restriction, among others. Macrophage migration
Yong-Chao Jiang et al.
Journal of biomedical materials research. Part A, 106(6), 1595-1603 (2018-02-11)
During the process of tissue regeneration facilitated by stem cells, physical properties of a scaffold affect behavior and activities of the cell. To enhance differentiation of human mesenchymal stem cells (MSCs) into endothelial-like cells (ELCs), we used electrospun fibrous substrates
Mi Jeong Kim et al.
Cellular signalling, 34, 110-120 (2017-03-23)
The nuclear factor kappa B (NF-κB) pathway is pivotal in controlling survival and apoptosis of cancer cells. Macrophage migration inhibitory factor (MIF), a cytokine that regulates the immune response and tumorigenesis under inflammatory conditions, is upregulated in various tumors. However
Yu Zhang et al.
Clinical and experimental pharmacology & physiology, 43(11), 1134-1144 (2016-10-21)
Macrophage migration inhibitory factor (MIF), a pleiotropic pro-inflammatory cytokine, is a key regulator in both innate and acquired immunity systems. MIF has become a promising drug target for inflammatory diseases. Apart from its cytokine activities, MIF is known to act

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service