Skip to Content
Merck
All Photos(1)

Key Documents

E6261

Sigma-Aldrich

β-Endorphin human

≥95% (HPLC)

Synonym(s):

β-Lipotropin 61-91, β-Lipotropin C-fragment, LPH (61-91)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C158H251N39O46S
CAS Number:
Molecular Weight:
3464.98
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](Cc3ccccc3)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc4ccc(O)cc4)[C@@H](C)O)[C@@H](C)O)C(C)C)[C@@H](C)O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc5ccc(O)cc5)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C158H251N39O46S/c1-17-84(9)126(153(237)182-102(44-29-34-65-163)137(221)186-112(74-118(166)206)142(226)171-86(11)131(215)183-110(73-94-48-52-96(204)53-49-94)146(230)177-99(41-26-31-62-160)135(219)175-98(40-25-30-61-159)134(218)170-78-122(210)173-106(158(242)243)56-59-124(213)214)193-154(238)127(85(10)18-2)192-132(216)87(12)172-143(227)113(75-119(167)207)185-136(220)100(42-27-32-63-161)178-147(231)111(72-92-38-23-20-24-39-92)184-144(228)107(68-81(3)4)188-155(239)129(89(14)201)195-152(236)125(83(7)8)191-148(232)108(69-82(5)6)187-151(235)116-45-35-66-197(116)157(241)130(90(15)202)196-140(224)103(54-57-117(165)205)179-149(233)114(79-198)189-138(222)101(43-28-33-64-162)176-139(223)104(55-58-123(211)212)180-150(234)115(80-199)190-156(240)128(88(13)200)194-141(225)105(60-67-244-16)181-145(229)109(71-91-36-21-19-22-37-91)174-121(209)77-168-120(208)76-169-133(217)97(164)70-93-46-50-95(203)51-47-93/h19-24,36-39,46-53,81-90,97-116,125-130,198-204H,17-18,25-35,40-45,54-80,159-164H2,1-16H3,(H2,165,205)(H2,166,206)(H2,167,207)(H,168,208)(H,169,217)(H,170,218)(H,171,226)(H,172,227)(H,173,210)(H,174,209)(H,175,219)(H,176,223)(H,177,230)(H,178,231)(H,179,233)(H,180,234)(H,181,229)(H,182,237)(H,183,215)(H,184,228)(H,185,220)(H,186,221)(H,187,235)(H,188,239)(H,189,222)(H,190,240)(H,191,232)(H,192,216)(H,193,238)(H,194,225)(H,195,236)(H,196,224)(H,211,212)(H,213,214)(H,242,243)/t84-,85-,86-,87-,88+,89+,90+,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,125-,126-,127-,128-,129-,130-/m0/s1

InChI key

JMHFFDIMOUKDCZ-NTXHZHDSSA-N

Gene Information

human ... POMC(5443)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-Tyr-Lys-Lys-Gly-Glu

General description

β-Endorphin is a potent endogenous opioid protein that is derived from propiomelanocortin, a protein found in the brain, anterior pituitary, skin, immune system, and other peripheral sites. β-endorphin, however, is localized to the brain and spinal cord. It is one of the major opioid peptides, and is the derivative of the C-terminal of β-lipotropin. It is made of 31 amino acids.

Application

Human β-Endorphin is suitable for-
  • radioimmunoassay to determine the portal blood β-endorphin levels in lactating ewes, and its relation to bouts of suckling
  • to assess the melanogenesis, proliferation and dendricity of melanocytes
  • to assess the expression of μ-opioid receptor, in human skin organ culture, on exposure to β-endorphin
  • to examine the effects on cardiovascular functions of β-endorphin injection nucleus in amygdaloideus centralis (AC), into either nucleus paraventricularis (NPV) or rostral ventrolateral medulla (RVL)

Biochem/physiol Actions

β-Endorphin acts as a ligand for both μ and δ opioid receptors in different parts of the brain. It is released in response to painful stimuli and has potent antinociceptive activity. When given supraspinally, this activity is mediated through its action on ε-opioid receptors in brain that results in the release of Met-enkephalin. This activity is also mediated by its interaction with δ2-opioid receptors in the spinal cord. When introduced intrathecally, this function is an outcome of its interaction with μ and κ-opioid receptors in the rat. In autoimmune disorders, the elevated production of cytokines results in the production of β-endorphin, in pituitary and lymphocytes. This acts as a negative regulator of antibody production, and is thus, implicated in autoimmune diseases.
Potent endogenous opioid protein that is derived from propiomelanocortin, a protein found in the brain, anterior pituitary, skin, immune system, and other peripheral sites. It is released in response to painful stimuli and has potent antinociceptive activity that is mediated through its action on μ receptors in brain and by μ and κ receptors in the spinal cord.
Potent endogenous opioid protein; released in response to painful stimuli and has potent antinociceptive activity.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bigliardi-Qi et al.
The Journal of investigative dermatology, 114(3), 527-532 (2000-02-26)
It has been reported that opioid peptides modulate the differentiation of normal human keratinocytes and that mu-opiate receptors are expressed in human epidermis. The regulation of keratinocyte differentiation is particularly important in psoriasis, and one of the markers for hyperproliferative
H Mørch et al.
Autoimmunity, 21(3), 161-171 (1995-01-01)
The immune system and the neuroendocrine system are closely interconnected having such means of bidirectional communication and regulation. In this review, a hypothesis is put forward regarding the possible role of beta-endorphins in the pathogenesis of autoimmune diseases: It is
Y H Ku et al.
Peptides, 22(9), 1465-1470 (2001-08-22)
It has been proved that input of specific electroacupuncture (EA) can activate beta-endorphin(beta-EP)ergic and noradrenergic neurons projecting to the rostral ventrolateral medulla (RVL), the latter acting upon the RVL-GABAergic interneurons, thereby produce depressor effect. The present study further shows that:
L F Tseng et al.
Naunyn-Schmiedeberg's archives of pharmacology, 351(5), 464-468 (1995-05-01)
The antinociception induced by beta-endorphin given supraspinally has been previously demonstrated to be mediated by the stimulation of epsilon-, but not mu-, delta- or kappa-opioid receptors in rats and mice. The present study was designed to determine what types of
K Gordon et al.
Journal of reproduction and fertility, 79(2), 397-408 (1987-03-01)
Matched hypothalamo-pituitary portal and jugular blood samples were collected over about 6 h from 7 lactating Corriedale ewes penned with their lambs, and a careful record was kept of ewe/lamb behaviour. Hypothalamo-pituitary portal blood concentrations of beta-endorphin were measured by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service