Skip to Content
Merck
All Photos(2)

Key Documents

E4267

Sigma-Aldrich

9-Ethylguanine

Synonym(s):

2-Amino-9-ethyl-6-hydroxypurine, 6-Amino-9-ethyl-2-hydroxypurine, 9-EtG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N5O
CAS Number:
Molecular Weight:
179.18
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥98%

Quality Level

form

powder

solubility

1 M NaOH: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

CCn1cnc2C(=O)N=C(N)Nc12

InChI

1S/C7H9N5O/c1-2-12-3-9-4-5(12)10-7(8)11-6(4)13/h3H,2H2,1H3,(H3,8,10,11,13)

InChI key

WDOYBEPLTCFIRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

9-Ethylguanine (9-EtG) is used as a DNA model base for a variety of physical chemical studies of organometallic complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna F A Peacock et al.
Journal of the American Chemical Society, 129(11), 3348-3357 (2007-02-27)
Potential biological and medical applications of organometallic complexes are hampered by a lack of knowledge of their aqueous solution chemistry. We show that the hydrolytic and aqueous solution chemistry of half-sandwich OsII arene complexes of the type [(eta6-arene)Os(XY)Cl] can be
Amanda P Neves et al.
Journal of inorganic biochemistry, 119, 54-64 (2012-11-29)
Several chlorido and amino Pt(2+) complexes of 2-hydroxy-3-(aminomethyl)-1,4-naphthoquinone Mannich bases HL exhibiting moderate to high cytotoxicity against cancer cell lines were studied in order to investigate their modes of DNA binding, in vitro DNA strand breaks, mechanism of topoisomerase (Topo
Zhe Liu et al.
Journal of medicinal chemistry, 54(8), 3011-3026 (2011-03-30)
The low-spin 5d(6) Ir(III) organometallic half-sandwich complexes [(η(5)-Cp(x))Ir(XY)Cl](0/+), Cp(x) = Cp*, tetramethyl(phenyl)cyclopentadienyl (Cp(xph)), or tetramethyl(biphenyl)cyclopentadienyl (Cp(xbiph)), XY = 1,10-phenanthroline (4-6), 2,2'-bipyridine (7-9), ethylenediamine (10 and 11), or picolinate (12-14), hydrolyze rapidly. Complexes with N,N-chelating ligands readily form adducts with 9-ethylguanine
Zhe Liu et al.
Inorganic chemistry, 50(12), 5777-5783 (2011-05-31)
Replacing the N,N-chelating ligand 2,2'-bipyridine (bpy) in the Ir(III) pentamethylcyclopentadienyl (Cp*) complex [(η(5)-C(5)Me(5))Ir(bpy)Cl](+) (1) with the C,N-chelating ligand 2-phenylpyridine (phpy) to give [(η(5)-C(5)Me(5))Ir(phpy)Cl] (2) switches on cytotoxicity toward A2780 human ovarian cancer cells (IC(50) values of >100 μM for 1
Flavia Barragán et al.
Journal of the American Chemical Society, 133(35), 14098-14108 (2011-07-30)
A photoactivated ruthenium(II) arene complex has been conjugated to two receptor-binding peptides, a dicarba analogue of octreotide and the Arg-Gly-Asp (RGD) tripeptide. These peptides can act as "tumor-targeting devices" since their receptors are overexpressed on the membranes of tumor cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service