Skip to Content
Merck
All Photos(2)

Key Documents

E2645

Sigma-Aldrich

Epidermal Growth Factor Receptor human

lyophilized powder, ≥15,000 units/mg protein (Bradford)

Synonym(s):

ERBB, ErbB1, HER1, EGFR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

form

lyophilized powder

potency

>15000.00 units/mg

specific activity

≥15,000 units/mg protein (Bradford)

mol wt

~170 kDa

packaging

pkg of 500UN

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell based assay: suitable

color

white

solubility

glycerol: 10%, clear to slightly hazy, colorless

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... EGFR(1956)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell signaling

Epidermal growth factor receptor (EGFR) belongs to a family of cell surface receptors called receptor tyrosine kinases (RTKs) and is also the most studied member of this family. The EGFR is encoded by the ErbB gene, mapped to chromosome 7 q22. These receptors are attached to the cytoplasmic membrane and consist of an ectodomain (ECD), comprising of four subdomains called DI-DIV, a hydrophobic transmembrane domain, and an intracytoplasmic tyrosine kinase domain.

Biochem/physiol Actions

170 kDa glycoprotein having EGF-activated protein tyrosine kinase activity; high levels of EGFR are expressed in approximately one third of human epithelial tumors, and in the cancers of the bladder, breast or lung that have poor clinical prognosis.
EGF exerts its actions by binding to the EGF receptor (EGFR), a 170 kDa glycoprotein having EGF-activated protein tyrosine kinase activity. Even in the absence of EGF-like ligands, EGFR can participate in cellular responses elicited by several other stimuli. High levels of EGFR are expressed in approximately one third of human epithelial tumors, and in the cancers of the bladder, breast or lung that have poor clinical prognosis, hence the interest in targeting EGFR.
Epidermal growth factor receptor (EGFR) has an essential role in normal developmental processes, and is mutated in multiple cancers. It is involved in the pathogenesis of epidermoid tumors, and thus, has extreme importance as a cancer therapeutic target. Gefitinib (GEF) and erlotinib act as EGFR inhibitors, and induce apoptosis in non-small cell lung cancer (NSCLC) cells. A truncated isoform, called EGFRvIII is linked to multiple cases of head and neck squamous cell carcinoma (HNSCC). In ∼40% of glioblastoma multiforme (GBM), this gene is amplified.

Unit Definition

One unit (U) of the enzyme is defined as the amount needed to incorporate 1 pmol of phosphate into the substrate (KVEKIGEGTYGVVYK: 6 - 20 residue of p34cdc2) in 1 minute.

Physical form

Lyophilized from a solution in 50 mM HEPES, pH 7.6, 150 mM NaCl, 0.05% Triton X-100, 1 mM dithiothreitol, and 10% trehalose.

Preparation Note

Affinity purified from human carcinoma A431 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Epidermal Growth Factor Receptor Pathway: A Model for Targeted Therapy
Scaltriti M and Baselga J
Journal of Separation Science null
A comprehensive pathway map of epidermal growth factor receptor signaling
Oda K, et al.
Molecular Systems Biology (2005)
Qian Gou et al.
Oncogene, 39(25), 4844-4853 (2020-05-23)
In response to nutrient deficiency, autophagy degrades cytoplasmic materials and organelles in lysosomes, which is nutrient recycling, whereas activation of EGFR mediates autophagy suppression in response to growth factors. It is unclear whether PPARδ could be the regulator of autophagy
EGFR/MDM2 signaling promotes NF-?B activation via PPAR? degradation
Xu Y, et al.
Carcinogenesis, 37(2), 215-222 (2016)
Epidermal Growth Factor Receptor Cell Proliferation Signaling Pathways
Wee P and Wang Z
Cancers, 9(5), 52-52 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service