Skip to Content
Merck
All Photos(1)

Key Documents

E1631

Sigma-Aldrich

β-Estradiol 17-valerate

≥98% (HPLC), powder

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 17-pentanote

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O3
CAS Number:
Molecular Weight:
356.50
Beilstein:
2480357
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Product Name

β-Estradiol 17-valerate, ≥98%

biological source

synthetic

Assay

≥98%

form

powder

technique(s)

ELISA: suitable

solubility

acetone: 25 mg/mL, hazy, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCC(=O)OC1CCC2C3CCc4cc(O)ccc4C3CCC12C

InChI

1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3

InChI key

RSEPBGGWRJCQGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Estradiol 17-valerate is a synthetic estrogen.

Application

β-Estradiol 17-valerate has been used:
  • in an in-vivo assay, to synchronize the estrus cycle in athymic nude mice, to study the effect of recombinant pigment epithelium-derived factor (rPEDF) on induced ectopic fibroid lesions
  • to study the influence of ovarian hormones on type II cGMP-dependent protein kinase (Prkg2) expression in mice
  • to induce growth of estrogen-dependent tumor cells in athymic nude mice to establish a breast cancer mouse model

Biochem/physiol Actions

Estradiol valerate is observed to be useful for treating menopausal climacteric symptoms. β-Estradiol 17-valerate is used as a source of 17β-estradiol (E2) in in vitro fertilization, hormone replacement therapy, contraceptive drugs, to study reproductive disorders in animal models. It serves as growth promoter in animal farming. β-Estradiol 17-valerate is also used to culture single-sex population among fish. Studies in rats show that β-Estradiol 17-valerate has negative effects on the estrus cycle pattern.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingli Lei et al.
Aquatic toxicology (Amsterdam, Netherlands), 134-135, 128-134 (2013-04-24)
β-estradiol 17-valerate (EV) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs and is detected in natural waters. Although EV is known as an estrogenic chemical, there is still a lack of
Marika Christina Möller et al.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology, 29(2), 173-176 (2012-10-26)
Both estrogen and testosterone insufficiency has been associated with reduced psychological well-being including fatigue. However, hormonal replacement studies on fatigue are rare. Therefore, we wanted to study the effect of testosterone and estrogen replacement therapy on cognitive fatigue and the
Elizabeth Micks et al.
Women's health (London, England), 7(5), 513-524 (2011-09-02)
Estradiol valerate and dienogest have been combined to create a novel four-phasic oral contraceptive pill effective for both pregnancy prevention and treatment of heavy menstrual bleeding. This formulation represents the only oral contraceptive pill available in the USA containing an
Gonzalo Cruz et al.
Fertility and sterility, 98(5), 1283-1290 (2012-08-03)
To investigate the developmental window in which E(2) exposure produces irreversible changes in ovarian function resulting in polycystic ovary. Basic experimental study. University animal laboratory. Thirty Sprague-Dawley rats were administered a single E(2) valerate dose (10 mg/kg of weight) at
L J Seal et al.
The Journal of clinical endocrinology and metabolism, 97(12), 4422-4428 (2012-10-12)
Breast development in transwomen is an important issue, affecting general psychological functioning. Current hormonal therapies are imperfect, with 60% of patients requesting mammoplasty. Interventions included the following: 1) comparing the effects on transwomen's requests for mammoplasty of estrogen valerate, ethinylestradiol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service