Skip to Content
Merck
All Photos(1)

Documents

E1270

Sigma-Aldrich

ECM Gel from Engelbreth-Holm-Swarm murine sarcoma

liquid, BioReagent, suitable for cell culture

Synonym(s):

Basement membrane extract (BME), EHS matrix

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product line

BioReagent

Quality Level

form

liquid

concentration

8-12 mg/mL

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Application

ECM gel can used with epithelial cells, endothelial cells, muscle cells, nerve cells and tumor cells.

Biochem/physiol Actions

The ECM gel will undergo thermally activated polymerization when brought to 20-40°C, forming a reconstituted basement membrane. PC12 cells will show neurite formation within 2 days when they are grown on a thin layer of this ECM gel.

Every mouse colony used for the production of this product is routinely screened for several pathogens. Tested and found negative for LDEV.

Components

ECM was prepared to a protein concentration of 8-12 mg/mL (in DMEM), containing laminin as a major component, collagen type IV, heparin sulfate proteoglycan, entactin, and other minor components. Addition of collagen type IV to the ECM gel increases polymerization.

Product contains 50 mg/L Gentamicin Sulfate.

Caution

For long term storage, keep product at -20°C. The ECM gel may be stored at 2-8°C for up to 72 hours.

Preparation Note

This product is prepared from Engelbreth-Holm-Swarm sarcoma in mice; dialyzed against chloroform. Gel should be thawed overnight at 2-8°C before use and dispensed to a multiwall plate using a plate and pipettes that are pre-cooled to 2-8°C. The gel may also be diluted up to twofold with 2-8°C Dulbecco′s Modified Eagle′s Medium, and should be done before gel is added to the plate. The product will gel within 5 minutes at 20°C. For prolonged manipulations, work should be conducted below 10°C. Cells can be plated on top of a thin gel layer of 0.5 mm or cultured inside a 1 mm layer. If you culture cells inside, add the cells to the gel prior to plating at a recommended density of 3-4 x 104 cells per mL. To dissociate cells from this gel, use protease in PBS without calcium, magnesium, or EDTA, at a concentration of 0.6-2.4 units per mL.

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lutian Yao et al.
Cytokine, 89, 194-200 (2016-02-13)
Pancreatic cancer is characterized as inflammatory malignancy with a dismal prognosis. There is abundant intratumoral infiltration of macrophages, and most of these tumor associated macrophages (TAM) are induced to be M2 phenotype. The M2 polarized TAM has been demonstrated to
Eva Lichtenegger et al.
Human gene therapy, 30(1), 44-56 (2018-06-20)
Muscle-invasive bladder cancer represents approximately 25% of diagnosed bladder cancer cases and carries a significant risk of death. Oncolytic viruses are novel antitumor agents with the ability to selectively replicate and lyse tumor cells while sparing healthy tissue. We explored
Jesper Just et al.
Scientific reports, 10(1), 5835-5835 (2020-04-05)
Ischemic exercise conducted as low-load blood flow restricted resistance exercise (BFRE) can lead to muscle remodelling and promote muscle growth, possibly through activation of muscle precursor cells. Cell activation can be triggered by blood borne extracellular vesicles (EVs) as these
Ievgenia Pastushenko et al.
Nature, 556(7702), 463-468 (2018-04-20)
In cancer, the epithelial-to-mesenchymal transition (EMT) is associated with tumour stemness, metastasis and resistance to therapy. It has recently been proposed that, rather than being a binary process, EMT occurs through distinct intermediate states. However, there is no direct in
Mallory E Walters et al.
Antioxidants (Basel, Switzerland), 9(6) (2020-07-02)
The aim of this work was to determine the physicochemical and biological activities of hydrolyzed proteins from sonicated oat brans. In addition to the control bran sample, two types of pre-treatment procedures-namely, ultrasonic bath and probe-type sonication-were performed to extract

Articles

Cell culture protocols using ECM Gel Matrix EHS basement membrane extract (BME) for neurite outgrowth, cell invasion and angiogenesis tube formation assays.

Cell culture protocols using ECM Gel Matrix EHS basement membrane extract (BME) for neurite outgrowth, cell invasion and angiogenesis tube formation assays.

Cell culture protocols using ECM Gel Matrix EHS basement membrane extract (BME) for neurite outgrowth, cell invasion and angiogenesis tube formation assays.

Cell culture protocols using ECM Gel Matrix EHS basement membrane extract (BME) for neurite outgrowth, cell invasion and angiogenesis tube formation assays.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service