Skip to Content
Merck
All Photos(4)

Key Documents

D9500

Sigma-Aldrich

2′-Deoxyguanosine 5′-monophosphate sodium salt hydrate

≥99% (HPLC)

Synonym(s):

5′-Deoxyguanylic acid, dGMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O7P · xNa+ · yH2O
Molecular Weight:
347.22 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O.[Na+].NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](COP(O)([O-])=O)O3)C(=O)N1

InChI

1S/C10H14N5O7P.Na.H2O/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(22-6)2-21-23(18,19)20;;/h3-6,16H,1-2H2,(H2,18,19,20)(H3,11,13,14,17);;1H2/q;+1;/p-1/t4-,5+,6+;;/m0../s1

InChI key

UVKXYXSKMTUDML-FVALZTRZSA-M

Related Categories

Application

2′-Deoxyguanosine 5′-monophosphate (dGMP) is used as a substrate of guanylate kinase(s) (EC 2.7.4.8) to form dGDP which upon phosphorylation to dGTP supports DNA biosynthesis. dGMP is use to study physiochemical characteristics of guanine based molecules.
2′-Deoxyguanosine 5′-monophosphate sodium salt hydrate has been used:
  • in reversed-phase high pressure liquid chromatography (HPLC) analysis[1]
  • to determine the effects of deoxy-mononucleotides on spontaneous proliferating CaCo-2 cell lines[2]
  • to determine its capacity to stimulate immune cell growth and function and used influenza virus antigen as an immunostimulant in vitro[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahmoud Kandeel et al.
Molecular and biochemical parasitology, 159(2), 130-133 (2008-04-01)
The present work describes cloning, expression, purification, characterization, and mutation of Plasmodium falciparum guanylate kinase (PlasmoDB ID PFI1420w). Amino-acid sequence alignment revealed important differences especially in K42-V51, Y73-A77, and F100-L110, which include residues important for kinase activity, and at helix
Brian G Gentry et al.
Biochemical pharmacology, 81(1), 43-49 (2010-09-18)
Many fraudulent nucleosides including the antivirals acyclovir (ACV) and ganciclovir (GCV) must be metabolized to triphosphates to be active. Cyclopropavir (CPV) is a newer, related guanosine nucleoside analog that is active against human cytomegalovirus (HCMV) in vitro and in vivo.
Ana-Maria Ruxandra Eftimie et al.
Roumanian archives of microbiology and immunology, 66(1-2), 22-25 (2008-10-22)
Guanylate kinase is a member of the nucleoside monophosphate (NMP) kinase family, a family of enzymes that despite having a low primary structure identity share a similar fold, which consists of three structurally distinct regions termed the CORE, LID, and
Dietary nucleotides and intestinal cell lines: I. Modulation of growth
Holen E and Jonsson R
Nutrition Research (New York, N.Y.), 24(3), 197-207 (2004)
Mahmoud Kandeel et al.
Journal of molecular recognition : JMR, 24(2), 322-332 (2011-03-02)
Plasmodium deoxyguanylate pathways are an attractive area of investigation for future metabolic and drug discovery studies due to their unique substrate specificities. We investigated the energetic contribution to guanylate kinase substrate binding and the forces underlying ligand recognition. In the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service