Skip to Content
Merck
All Photos(1)

Key Documents

D7408

Sigma-Aldrich

2′,5′-Dideoxyadenosine

≥95% (HPLC), solid

Synonym(s):

2ʹ,5ʹ-dd-Ado, NSC 95943

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O2
CAS Number:
Molecular Weight:
235.24
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

C[C@H]1O[C@H](C[C@@H]1O)n2cnc3c(N)ncnc23

InChI

1S/C10H13N5O2/c1-5-6(16)2-7(17-5)15-4-14-8-9(11)12-3-13-10(8)15/h3-7,16H,2H2,1H3,(H2,11,12,13)/t5-,6+,7-/m1/s1

InChI key

FFHPXOJTVQDVMO-DSYKOEDSSA-N

Gene Information

Application

2′,5′-Dideoxyadenosine has been used to elucidate the mechanism of diligustilide (DLG). It has also been used to inhibit adenylate cyclase (AC).

Biochem/physiol Actions

Cell-permeable adenylyl cyclase inhibitor. IC50 = 2.7 μM in detergent-dispersed rat brain preparations.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Reconstitution

Store at −20 °C after reconstitution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fen Hu et al.
Molecular medicine reports, 24(4) (2021-07-31)
Inflammation and oxidative stress have indispensable roles in the development of acute lung injury (ALI). MicroRNA (miRNA/miR)‑351‑5p was initially identified as a myogenesis‑associated miRNA; however, its role in lipopolysaccharide (LPS)‑induced ALI remains unclear. The aim of the present study was
Rula Azzam et al.
AIDS research and human retroviruses, 22(7), 619-629 (2006-07-13)
HIV-1 infection of cells of macrophage lineage impairs a number of effector functions performed by these cells, including phagocytosis of opsonized pathogens. In this study we investigate the effects of HIV-1 on the mechanism of complement (C')-mediated phagocytosis by human
Hisatoshi Sugiura et al.
Journal of cellular physiology, 210(1), 99-110 (2006-09-26)
Prostaglandin E(2) (PGE(2)) has been shown to have a strong cytoprotective effect, inhibiting apoptosis. In the present study, we evaluated whether PGE(2) has a protective effect on cigarette smoke extract (CSE)-induced apoptosis in human lung fibroblasts. Apoptosis was assessed by
Zhixiong Chen et al.
International journal for parasitology, 37(7), 743-761 (2007-02-20)
Trichinella spiralis infection causes hyperexcitability in enteric after-hyperpolarising (AH) sensory neurons that is mimicked by neural, immune or inflammatory mediators known to stimulate adenylyl cyclase (AC)/cyclic 3',5'-adenosine monophosphate (cAMP) signaling. The hypothesis was tested that ongoing modulation and sustained amplification
Jorge A Rodriguez et al.
American journal of physiology. Heart and circulatory physiology, 290(1), H30-H36 (2005-09-07)
Vascular smooth muscle cell proliferation and migration play an important role in the pathophysiology of several vascular diseases, including atherosclerosis. Prostaglandins that have been implicated in this process are synthesized by two isoforms of cyclooxygenase (COX), with the expression of

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service