Skip to Content
Merck
All Photos(1)

Key Documents

D4168

Sigma-Aldrich

SIGMAFAST 3,3′-Diaminobenzidine tablets

SIGMAFAST, tablet

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 1 mL

Quality Level

form

tablet

solubility

deionized water: 1 mL, clear, pink

storage temp.

−20°C

SMILES string

Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Developed for use in immunohistology and immunoblotting as a precipitating substrate for the localization of peroxidase activity. DAB is the immunohistology substrate of choice as it produces an intense brown-black stain which is resistant to alcohol. Slides stained with DAB may be dehydrated, mounted in resinous media, and stored for future reference. SIGMAFAST DAB Tablets require no additional buffers or steps to prepare an active substrate solution. Tablets are individually packaged in foil packets

Reconstitution

Each tablet set dissolved in 1 mL deionized water yields a ready-to-use buffered solution containing DAB and urea hydrogen peroxide.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Ox. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

5.1B - Oxidizing hazardous materials

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin M Byers et al.
ACS omega, 5(9), 4673-4681 (2020-03-17)
Two-dimensional paper networks (2DPNs) have enabled the use of paper-based platforms to perform multistep immunoassays for detection of pathogenic diseases at the point-of-care. To date, however, detection has required the user to provide multiple signal enhancement solutions and been limited
Soner Celik et al.
Journal of assisted reproduction and genetics, 35(4), 615-626 (2018-03-03)
Even with 86 live births reported globally so far, the mechanism of primordial follicle loss following autotransplantation of the frozen-thawed ovarian tissue needs further evaluation. Pten, Tsc1, p27, and Amh are the inhibitor proteins that play crucial roles in suppressing
D Randazzo et al.
American journal of physiology. Cell physiology, 312(1), C16-C28 (2016-10-28)
We recently reported that skeletal muscle fibers of obscurin knockout (KO) mice present altered distribution of ankyrin B (ankB), disorganization of the subsarcolemmal microtubules, and reduced localization of dystrophin at costameres. In addition, these mice have impaired running endurance and
Itamar Goren et al.
Nitric oxide : biology and chemistry, 87, 31-42 (2019-03-14)
The gaseous mediators nitric oxide (NO), carbon monoxide (CO) and lately also hydrogen sulfide (H2S) have been described to contribute to the interplay of protein type- and lipid mediators in the regulation of wound healing. In particular, the recently reported
Dileyra Adiguzel et al.
PloS one, 14(5), e0216814-e0216814 (2019-05-24)
Recent studies indicate that FoxO1 has roles in female reproductive system, especially in maternal endometrium. Although various cellular aspects and molecular pathways have been identified, the exact molecular characteristics of embryo implantation are still not completely understood. In this study

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service