Skip to Content
Merck
All Photos(1)

Documents

D4071

Sigma-Aldrich

5, 15-DPP

Synonym(s):

5,15-Diphenyl-21H,23H-porphine, 5,15-Diphenylporphyrin, 5,15-diphenyl-Porphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N4
CAS Number:
Molecular Weight:
462.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

SMILES string

c1ccc(cc1)-c2c3ccc(cc4ccc([nH]4)c(-c5ccccc5)c6ccc(cc7ccc2[nH]7)n6)n3

InChI

1S/C32H22N4/c1-3-7-21(8-4-1)31-27-15-11-23(33-27)19-25-13-17-29(35-25)32(22-9-5-2-6-10-22)30-18-14-26(36-30)20-24-12-16-28(31)34-24/h1-20,33,36H/b23-19-,24-20-,25-19-,26-20-,31-27-,31-28-,32-29-,32-30-

InChI key

FNNOEKVUXXVPAI-DUVOQLPESA-N

Application

5, 15-DPP has been used as a signal transducer and activator of transcription 3 (STAT3) inhibitor:
  • to analyze its effects on malignant peripheral nerve sheath tumor (MPNST) survival and proliferation
  • to study its effects on the thymic stromal lymphopoietin (TSLP) modulation of nometastatic gene 23-H1 (NME1 or nm23-H1) expression in human trophoblasts
  • to study its effects on 17β-estradiol and 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD)-induced M2 polarization in human macrophages and endometrial stromal cells

Biochem/physiol Actions

5, 15-DPP is a selective STAT3 inhibitor and a selective STAT3-SH2 antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wang et al.
Clinical immunology (Orlando, Fla.), 143(1), 88-95 (2012-03-01)
Our previous study has demonstrated that TSLP induces the invasion of human trophoblasts and plays a role in embryo implantation. The present study is undertaken to explore the mechanism by which TSLP modulates trophoblast invasion. It was revealed that TSLP
Yun Wang et al.
PloS one, 10(5), e0125559-e0125559 (2015-05-08)
The goal of this study is to elucidate the effects of 17β-estradiol and TCDD (2,3,7,8-tetrachlorodibenzo-p-dioxin) on macrophage phenotypes in the endometriotic milieu. Co-culture of endometrial stromal cells (ESCs) and U937 cells (macrophage cell line) was performed to simulate the endometriotic
Jody Fromm Longo et al.
Cell communication and signaling : CCS, 17(1), 74-74 (2019-07-12)
We have found that erbB receptor tyrosine kinases drive Ras hyperactivation and growth in NF1-null malignant peripheral nerve sheath tumors (MPNSTs). However, MPNSTs variably express multiple erbB receptors with distinct functional characteristics and it is not clear which of these
Shaoxue Zeng et al.
Theranostics, 12(15), 6705-6722 (2022-10-04)
Rationale: Müller cells play an essential role in maintaining the health of retinal photoreceptors. Dysfunction of stressed Müller cells often results in photoreceptor degeneration. However, how these cells communicate under stress and the signalling pathways involved remain unclear. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service