D3659
cis-7,10,13,16-Docosatetraenoic acid
≥98% (GC)
Synonym(s):
Adrenic Acid, DTA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H36O2
CAS Number:
Molecular Weight:
332.52
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥98% (GC)
form
liquid
functional group
carboxylic acid
lipid type
omega FAs
shipped in
ambient
storage temp.
−20°C
SMILES string
CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCCCC(O)=O
InChI
1S/C22H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-21H2,1H3,(H,23,24)/b7-6-,10-9-,13-12-,16-15-
InChI key
TWSWSIQAPQLDBP-DOFZRALJSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Sealed ampule
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Thierry Durand et al.
Biochimie, 95(1), 86-90 (2012-09-27)
Oxidative damage has been reported in Rett syndrome (RTT), a pervasive development disorder mainly caused up to 95% of cases by mutations in the X-linked methyl-CpG binding protein 2 (MeCP2) gene. We have recently synthesized F(2)-Dihomo-isoprostanes (F(2)-Dihomo-IsoP), peroxidation products from
Carlos Guijas et al.
Biochimica et biophysica acta, 1821(11), 1386-1393 (2012-07-25)
Cells metabolize arachidonic acid (AA) to adrenic acid (AdA) via 2-carbon elongation reactions. Like AA, AdA can be converted into multiple oxygenated metabolites, with important roles in various physiological and pathophysiological processes. However, in contrast to AA, there is virtually
Martin C Schumacher et al.
Urologic oncology, 31(3), 318-324 (2011-03-19)
To analyze different polyunsaturated fatty acid (PUFA) tissue levels in malignant compared with benign prostatic tissue from the same prostate specimens. Fresh frozen benign and malignant prostatic tissue was obtained from radical prostatectomy specimens in 49 men with pathologic stage
T Hinckley et al.
Biology of reproduction, 55(2), 445-449 (1996-08-01)
Experiments were conducted to determine the effects of exogenous polyunsaturated fatty acids (PUFA) on the production of progesterone and prostanoids by dispersed bovine luteal cells and to characterize endogenous luteal fatty acids throughout the estrous cycle. The addition of eicosapentaenoic
E Christensen et al.
Scandinavian journal of clinical and laboratory investigation. Supplementum, 215, 61-74 (1993-01-01)
The metabolism of the C22 unsaturated fatty acids erucic acid (22:1(n-9)), adrenic acid (22:4(n-6)), docosapentaenoic acid (22:5(n-3)) and docosahexaenoic acid (22:6(n-3)) was studied in cultured fibroblasts from patients with acyl-CoA oxidase deficiency, the Zellweger syndrome, X-linked adrenoleukodystrophy (X-ALD) and normal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service