Skip to Content
Merck
All Photos(2)

Key Documents

D153

Sigma-Aldrich

(R)(−)-DOI hydrochloride

≥98% (HPLC), solid

Synonym(s):

(−)-1-(2,5-Dimethoxy-4-iodophenyl)-2-aminopropane hydrochloride, (−)-2,5-Dimethoxy-4-iodoamphetamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16INO2 · HCl
CAS Number:
Molecular Weight:
357.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]22/D −12.36°, c = 2 in H2O(lit.)

drug control

Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

solubility

H2O: ≥20 mg/mL
ethanol: soluble

SMILES string

Cl.COc1cc(C[C@@H](C)N)c(OC)cc1I

InChI

1S/C11H16INO2.ClH/c1-7(13)4-8-5-11(15-3)9(12)6-10(8)14-2;/h5-7H,4,13H2,1-3H3;1H/t7-;/m1./s1

InChI key

QVFDMWGKHUFODK-OGFXRTJISA-N

Gene Information

Biochem/physiol Actions

Potent and selective 5-HT2 serotonin receptor agonist that crosses the blood-brain barrier; more potent enantiomer of ±-DOI hydrochloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ishier Raote et al.
Molecular pharmacology, 83(1), 42-50 (2012-10-05)
G protein-coupled receptor (GPCR) signaling is modulated by endocytosis and endosomal sorting of receptors between degradation and recycling. Differential regulation of these processes by endogenous ligands and synthetic drugs is a poorly understood area of GPCR signaling. Here, we describe
M I Zaĭchenko et al.
Zhurnal vysshei nervnoi deiatelnosti imeni I P Pavlova, 63(2), 246-255 (2013-07-23)
The influence of drugs, agonist (DOI) and antagonist (ketanserin) ofserotonin receptors 5-HT2 on the behavior of rats tested by the method of choice to the value of reinforcement was investigated. Depending on their preferences in food reinforcement rats were divided
João Paulo Capela et al.
Neurotoxicology, 34, 254-263 (2012-09-18)
3,4-Methylenedioxymethamphetamine (MDMA or "Ecstasy") and 2,5-dimethoxy-4-iodoamphetamine hydrochloride (DOI) are hallucinogenic amphetamines with addictive properties. The hippocampus is involved in learning and memory and seems particularly vulnerable to amphetamine's neurotoxicity. We evaluated the neurotoxicity of DOI and MDMA in primary neuronal
Lena Wischhof et al.
Pharmacology, biochemistry, and behavior, 102(1), 6-12 (2012-04-04)
Prepulse inhibition (PPI) of the acoustic startle response (ASR) provides a measure of sensorimotor gating mechanisms that are impaired in schizophrenia patients. Interactions of the serotonin (5-hydroxytryptamine, 5-HT) and glutamatergic systems, especially via the 5-HT(2A) receptor subtype, have been implicated
[125I]-2-(2,5-Dimethoxy-4-iodophenyl)aminoethane ([125I]-2C-I) as a label for the 5-HT2 receptor in rat frontal cortex.
Johnson, et al.
Pharmacology, Biochemistry, and Behavior, 35, 211-211 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service