Skip to Content
Merck
All Photos(1)

Key Documents

D129

Sigma-Aldrich

R-(+)-DIOA

≥98% (HPLC), solid

Synonym(s):

R(+)-Butylindazone, R-(+)-[(2-n-Butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1H-inden-5-yl)oxy]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24Cl2O4
CAS Number:
Molecular Weight:
399.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]27/D +18.1°, c = 0.7 in methanol(lit.)

color

white

solubility

H2O: insoluble <0.11 mg/mL
DMSO: >20 mg/mL
0.1 M HCl: insoluble
0.1 M NaOH: soluble
ethanol: soluble

SMILES string

CCCC[C@]1(Cc2cc(OCC(O)=O)c(Cl)c(Cl)c2C1=O)C3CCCC3

InChI

1S/C20H24Cl2O4/c1-2-3-8-20(13-6-4-5-7-13)10-12-9-14(26-11-15(23)24)17(21)18(22)16(12)19(20)25/h9,13H,2-8,10-11H2,1H3,(H,23,24)/t20-/m0/s1

InChI key

YAWWQIFONIPBKT-FQEVSTJZSA-N

Application

R-(+)-DIOA has been used as a potassium/chloride K+-Cl- transport inhibitor to study its effects on:
  • short circuit current (Isc) in rat semicircular canal duct (SCCD) epithelium
  • uptake of grepafloxacin by THP-1 monocytes
  • the uptake of moxifloxacin by THP-1 monocytes

Biochem/physiol Actions

[(dihydroindenyl)oxy]acetic acid (DIOA) acts as a potent inhibitor of potassium/chloride (K+–Cl) co-transporter. It does not exert its effects on bumetanimide-sensitive [Na+,K+,Cl-]-cotransport system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R P Garay et al.
Naunyn-Schmiedeberg's archives of pharmacology, 334(2), 202-209 (1986-10-01)
3-Amino-5-sulfamoylbenzoic acids and several series of (aryloxy)alkanoic acids were evaluated for their inhibitory effects on two human erythrocyte ion transport systems--the Na+,K+ cotransport system and the DIDS-sensitive anion carrier. Several classic loop diuretics, including the (aryloxy)alkanoic acid-ethacrynic acid and several
The inhibition of Na(+)-Pi cotransport and Pi self-exchange mechanisms in lactating rat mammary tissue.
J M Shillingford et al.
Biochemical Society transactions, 23(4), 559S-559S (1995-11-01)
L D Ochoa-de la Paz et al.
Pflugers Archiv : European journal of physiology, 449(6), 526-536 (2005-01-05)
An increase in extracellular KCl ([KCl]o) occurs under various pathological conditions in the retina, leading to retinal swelling and possible neuronal damage. The mechanisms of this KCl o-induced retinal swelling were investigated in the present study, with emphasis on the
Tatsuya Asai et al.
The European journal of neuroscience, 15(11), 1737-1746 (2002-06-26)
With repetitive electrical stimulation of the dorsal root (20 Hz for 1 s at C-fibre strength), intrinsic optical signals (IOSs), measured as changes in light transmittance, were recorded in the superficial dorsal horn of rat spinal cord slices using a
Satyanarayana R Pondugula et al.
BMC physiology, 13, 6-6 (2013-03-30)
The vestibular system controls the ion composition of its luminal fluid through several epithelial cell transport mechanisms under hormonal regulation. The semicircular canal duct (SCCD) epithelium has been shown to secrete Cl- under β2-adrenergic stimulation. In the current study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service