Skip to Content
Merck
All Photos(1)

Key Documents

D0754

Sigma-Aldrich

3,5-Diiodo-L-tyrosine dihydrate

≥98.0% (HPLC), suitable for ligand binding assays

Synonym(s):

3,5-diiodo-L-tyrosine hydrate (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9I2NO3 · 2H2O
CAS Number:
Molecular Weight:
469.01
Beilstein:
2218691
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

3,5-Diiodo-L-tyrosine dihydrate, crystalline

Assay

≥98.0% (HPLC)

form

crystalline

technique(s)

ligand binding assay: suitable

color

white

mp

200 °C (dec.) (lit.)

solubility

4 M NH4OH in methanol: 50 mg/mL, clear, yellow-green

storage temp.

−20°C

SMILES string

N[C@@H](Cc1cc(I)c(O)c(I)c1)C(O)=O

InChI

1S/C9H9I2NO3/c10-5-1-4(2-6(11)8(5)13)3-7(12)9(14)15/h1-2,7,13H,3,12H2,(H,14,15)/t7-/m0/s1

InChI key

NYPYHUZRZVSYKL-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Substrate for the assay of halogenated tyrosine and thyroid hormone aminotransferase. Intermediate in the biosynthesis and alternative pathways of metabolism of thyroid hormones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Frýdlová et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 863(1), 135-140 (2008-02-08)
The interaction of porcine pepsin A with immobilized derivatives of aromatic amino acids was investigated. Divinyl sulfone-activated Sepharose was used to immobilize N-acetyl-l-phenylalanine and 3,5-diiodo-l-tyrosine via their free carboxyl groups and l-tyrosine via its amino group. Immobilized l-tyrosine was iodinated
Yining Zheng et al.
Talanta, 69(1), 107-112 (2008-10-31)
In this paper, a new method for separation, identification and quantitation of iodotyrosines and iodothyronines [3-monoiodo-L-tyrosine (MIT), 3,5-diiodo-L-tyrosine (DIT), L-thyronine (T(0)), 3,5-diiodo-L-thyronine (T(2)), 3,5,3'-triiodo-L-thyronine (T(3)) and 3,3',5,5'-tetraiodo-L-thyronine (T(4))] was described by using capillary electrophoresis with photodiode-array ultraviolet-visible detection (CE-UV). The
Sangeeta Kumari et al.
Journal of the American Society for Mass Spectrometry, 18(8), 1516-1524 (2007-06-26)
L-Tyrosine and iodinated L-tyrosines, i.e., 3-iodo-L-tyrosine and 3,5-diiodo-L-tyrosine, are successfully used as chiral references for the chiral discrimination of aliphatic, acidic, and aromatic amino acids. Chiral discrimination is achieved by investigating the collision-induced dissociation spectra of the trimeric complex [Cu(II)(ref)(2)(A)
M. Nakano
Methods in Enzymology, 17A, 660-660 (1970)
A Balsam et al.
The Journal of clinical investigation, 72(4), 1234-1245 (1983-10-01)
Studies were performed to elucidate the nature of the pathway of hepatic thyroxine (T4) metabolism that is activated by inhibitors of liver catalase. For this purpose, the metabolism of T4 in homogenates of rat liver was monitored with T4 labeled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service