Skip to Content
Merck
All Photos(2)

Key Documents

D0184

Sigma-Aldrich

2′-Deoxyuridine 5′-triphosphate sodium salt solution

100 mM in H2O

Synonym(s):

dUTP sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N2O14P3 · xNa+
CAS Number:
Molecular Weight:
468.14 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.52

Assay

≥99%

form

liquid

concentration

100 mM in H2O

color

colorless

foreign activity

DNase, RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].OC1CC(OC1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)N2C=CC(=O)NC2=O

InChI

1S/C9H15N2O14P3.Na.H/c12-5-3-8(11-2-1-7(13)10-9(11)14)23-6(5)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;/h1-2,5-6,8,12H,3-4H2,(H,18,19)(H,20,21)(H,10,13,14)(H2,15,16,17);;

InChI key

FVFWWPFKJNTHIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′-Deoxyuridine 5′-triphosphate (dUTP) is incorporated into DNA during DNA synthesis. It is one of the main intermediate during pyrimidine nucleotide biosynthesis. It is one of the sources of uracil in DNA. Deoxyuridine 5′-triphosphate nucleotidohydrolase suppresses the incorporation of dUTP into DNA. It converts dUTP to 2′-deoxyuridine 5′-monophosphate (dUMP).

Application

2′-Deoxyuridine 5′-triphosphate sodium salt solution has been used in the mouse genomic DNA amplification by polymerase chain reaction (PCR). It is also suitable for conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

dUTP incorporation into genomic DNA is linked to transcription in yeast.
Kim N
Nature, 459(7250), 1150-1153 (2009)
Abundant non-canonical dUTP found in primary human macrophages drives its frequent incorporation by HIV-1 reverse transcriptase.
Kennedy EM
The Journal of Biological Chemistry, 286(28), 25047-25055 (2011)
M C Longo et al.
Gene, 93(1), 125-128 (1990-09-01)
Polymerase chain reactions (PCRs) synthesize abundant amplification products. Contamination of new PCRs with trace amounts of these products, called carry-over contamination, yields false positive results. Carry-over contamination from some previous PCR can be a significant problem, due both to the
DNA N-glycosidases: properties of uracil-DNA glycosidase from Escherichia coli.
T Lindahl et al.
The Journal of biological chemistry, 252(10), 3286-3294 (1977-05-25)
Yan Jiang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(21), 7152-7167 (2010-05-28)
Histone methyltransferases specific for the histone H3-lysine 9 residue, including Setdb1 (Set domain, bifurcated 1)/Eset/Kmt1e are associated with repressive chromatin remodeling and expressed in adult brain, but potential effects on neuronal function and behavior remain unexplored. Here, we report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service