CLS431344
Corning® Roller Bottles, CellBIND® Cell Culture Surface
clear polystyrene, sterile, bottle surface area 850 cm2, cap (Easy Grip), case of 44
Synonym(s):
Corning roller bottles, roller bottles
About This Item
Recommended Products
material
cap (Easy Grip)
clear polystyrene
cylindrical bottle
high-density polyethylene cap
polystyrene bottle
wide-mouth bottle
sterility
sterile
sterile
feature
cap
disposable
graduations
packaging
case of 44
manufacturer/tradename
Corning 431344
bottle neck diam.
~45 mm
bottle surface area
850 cm2
bottle working volume
170-255 mL
capacity
255 mL
diam. × H
117 mm × 271 mm
binding type
CellBIND® surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- The Corning CellBIND cell culture surface
- Improves cell attachment leading to increased cell growth and yields
- Results in more consistent and even cell attachment
- Helps adapt cells to reduced serum or serum-free conditions
- Reduces premature cell detachment from confluent cultures
Features and Benefits
- Easy Grip Caps feature large knurls designed for ergonomic handling
- Easy Grip Vent Caps are designed for applications requiring consistent gas exchange
- Bottles are manufactured from virgin polystyrene, caps from high density polyethylene
- Both resins meet the USP Class VI requirements for plastic containers and closures
- Treated with a Corning CellBIND surface treatment for enhanced cell attachment
- One piece seamless construction with printed graduations
- Printed lot numbers to aid in product traceability
- Sterilized by g−radiation and certified non-pyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service