Key Documents
CLS4082
Corning® Lambda® plus 8-channel pipettor
volume 20-200 μL
Synonym(s):
Multi-channel pipette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
8-channel
manufacturer/tradename
Corning 4082
volume
20-200 μL
Related Categories
General description
  • Corning® Lambda® plus 8-channel pipettor are designed for universal fit with all pipet tips of common brands.
  • Adjustable tip ejector.
  • For volume 20-200 μL.
  • Thumb wheel or a plunger button to set volume
  • Smooth plunger movement and extremely low pipetting forces.
  • Accuracy (%) ±3.0 to ±1.0 and Precision (%) <1.5 to <0.6.
  • 3-year warranty and Factory calibrated to strict ISO 8655 standards.
  • Fully autoclavable and UV resistant.
  • Inclusive of a key for easy in-house calibration.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)