CLS353219
Corning® Falcon® Microplate
wells, 96, Tissue Culture (TC)-treated surface, flat bottom clear wells
Synonym(s):
Microplate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear wells
size:
96 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
wells 96
wells 96
material
flat bottom clear wells
sterility
sterile
feature
lid
wells 96
packaging
case of 32 ea (4 packs of 8)
manufacturer/tradename
Corning 353219
size
96 well
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Falcon® 96 Well Black with Clear Flat Bottom TC-Treated Imaging Plate with Lid, 8/Pack, 32/Case
Application
Assay
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service