CL6B200
Sepharose™ CL-6B Size Exclusion Resin
Cross-linked, 90-350 mesh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
Sepharose™ CL-6B, Cross-linked
form
suspension
bead diameter
40-165 μm
pore size
10,000-1,000,000 fractionation range (Dextrans)
10,000-4,000,000 fractionation range (Globular proteins)
storage temp.
2-8°C
Related Categories
General description
Sepharose CL is a cross-linked derivative of sepharose, prepared by reacting sepharose with 2,3-dibromopropanol under strongly alkaline conditions.
Application
Sepharose CL-6B is used in affinity chromatography, protein chromatography, gel filtration chromatography, separation media and resins. Sepharose™ CL-6B has been used to study in vitro inhibition of human melanoma cells and to gather data in other valuable anticancer studies.
Sepharose™CL-6B has been used for the purification of glucoamylase enzyme and targeted ribonucleoprotein (RNP).
Legal Information
Sepharose is a trademark of Cytiva
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
W Chen et al.
Journal of bacteriology, 175(17), 5677-5682 (1993-09-01)
A membrane-associated ATPase with an M(r) of approximately 510,000 and containing subunits with M(r)s of 80,000 (alpha), 55,000 (beta), and 25,000 (gamma) was isolated from the methanogen Methanococcus voltae. Enzymatic activity was not affected by vanadate or azide, inhibitors of
M Schweizer et al.
Molecular & general genetics : MGG, 203(3), 479-486 (1986-06-01)
FAS1, the structural gene of the pentafunctional fatty acid synthetase subunit beta in Saccharomyces cerevisiae has been sequenced. Its reading frame represents an intron-free nucleotide sequence of 5,535 base pairs, corresponding to a protein of 1,845 amino acids with a
Ru-Ping Zhang et al.
Sheng wu hua xue yu sheng wu wu li xue bao Acta biochimica et biophysica Sinica, 30(6), 561-564 (2002-08-09)
Luffin B, a plan single-chain ribosome inactivating protein, was purified from seeds of Luffa cylindrica by Blue Sepharose CL-6B affinity chromatography. An immunotoxin was constructed with luffin B and Ng76, a monoclonal antibody to human melanoma cell M(21). Luffin B-Ng76
D A Grahame
The Journal of biological chemistry, 266(33), 22227-22233 (1991-11-25)
An enzyme complex containing carbon monoxide dehydrogenase and a corrinoid protein has been isolated from Methanosarcina barkeri. Sodium dodecyl sulfate-gel electrophoresis revealed five polypeptides of molecular masses alpha = 19,700, beta = 84,500, gamma = 63,200, delta = 53,000, and
Amylolytic fungi in starter cakes for rice beer production
Das AJ, et al.
The Journal of General and Applied Microbiology, 63(4), 236-245 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service