IPI-2I
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Porcine intestine
packaging
tube of 5 μg 93100622-DNA-5UG
pkg of vial of cells 93100622-1VL
growth mode
Adherent
karyotype
Heteroploid
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Boar Miniature male ileum, SV40 transformed
Cell Line Description
Epitheloid cell line derived from the ileum of an adult histocompatible miniature male boar. Transformed by SV40 plasmid pSV3-neo. Electron microscopy analysis shows evidence of microvilli and desmosomes connecting cells. The cells were found to co-express cytokeratin 18 specific of simple luminal epithelial cells and vimentin associated with a mesenchymal origin. Susceptible to Transmissible Gastro-enteritis (TG) virus.
Application
Studies of epithelial cell differentiation.
Culture Medium
DMEM + 4mM Glutamine + 0.024IU/ml Insulin + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service