AFRC MAC 57
86030310, Reactivity: Anti-Lipopolysaccharide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Rat x Rat Hybridoma
ImmunoDon
LOU/Iap rat spleen
Myeloma
Y3Ag1.2.3
Reactivity
Anti-Lipopolysaccharide
technique(s)
cell culture | mammalian: suitable
isotype
IgM
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat Y3Ag1.2.3 x Rat LOU/Iap spleen
Cell Line Description
The antibody is directed against lipopolysaccharide components of the bacterial cell wall and also with lipopolysaccharide from free-living cultures of Rhizobium strains 3841 and B155.
Immunogen
Sonicated bacteroid material from Rhizobium leguminosarum strain 3841 and strain B155
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service