Skip to Content
Merck
All Photos(1)

Documents

C8788

Sigma-Aldrich

Complement C3 deficient serum human

substrate serum for C3 activity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Quality Level

form

solution

usage

substrate serum for C3 activity

technique(s)

activity assay: suitable

impurities

infectious agent, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C3(718)

General description

Complement C3 deficient serum is a normal human serum in which C3 protein is removed by affinity chromatography. Depleted serum helps in the complement cascade up to the preferred convertase level. It eases the formation of enzyme complex from purified components.

Application

Complement C3 deficient serum human has been used:
  • as a component of the culture medium to study the influence of the inhibition of complement activation on complement receptors (CR3 and C3aR) expression in CD3+CD56+NKT-like cells
  • as a negative control to evaluate the downstream complement activation post-mannan-binding lectin (MBL) pathway activation
  • to study the opsonization of foreign particles by complement system

Physical form

Supplied as a solution in phosphate buffered saline, pH 7.4

Analysis Note

C3 is depleted by immunoadsorption as judged by a highly sensitive hemolytic assay.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Celik et al.
Infection and immunity, 69(12), 7304-7309 (2001-11-14)
The complement system and the natural antibody repertoire provide a critical first-line defense against infection. The binding of natural antibodies to microbial surfaces opsonizes invading microorganisms and activates complement via the classical pathway. Both defense systems cooperate within the innate
Marcin Okroj et al.
PloS one, 7(10), e47245-e47245 (2012-10-17)
C3 and C5 convertases are central stages of the complement cascade since they converge the different initiation pathways, augment complement activation by an amplification loop and lead to a common terminal pathway resulting in the formation of the membrane attack
Shin Yong Park et al.
Journal of immunology (Baltimore, Md. : 1950), 181(9), 6328-6336 (2008-10-23)
Enterococcus faecalis (Ef) accounts for most cases of enterococcal bacteremia, which is one of the principal causes of nosocomial bloodstream infections (BSI). Among several virulence factors associated with the pathogenesis of Ef, an extracellular gelatinase (GelE) has been known to
Xiao-Yun Min et al.
Human immunology, 82(9), 625-633 (2021-06-18)
CD3+CD56+NKT-like cells are a rare population of lymphocytes that serve important roles in various types of immune-related diseases, and particularly in cancer. The complement system regulates inflammatory and immune responses by interacting with complement receptors expressed on a range of
Xiao-Yun Min et al.
Human immunology, 82(9), 625-633 (2021-06-18)
CD3+CD56+NKT-like cells are a rare population of lymphocytes that serve important roles in various types of immune-related diseases, and particularly in cancer. The complement system regulates inflammatory and immune responses by interacting with complement receptors expressed on a range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service