Skip to Content
Merck
All Photos(1)

Key Documents

C8124

Sigma-Aldrich

CH-223191

≥98% (HPLC), powder, AhR antagonist

Synonym(s):

1-Methyl-N-[2-methyl-4-[2-(2-methylphenyl)diazenyl]phen yl-1H-pyrazole-5-carboxamide, 2-Methyl-2H-pyrazole-3-carboxylic acid (2-methyl-4-o-tolylazo-phenyl)-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N5O
CAS Number:
Molecular Weight:
333.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

CH-223191,

Assay

≥98% (HPLC)

Quality Level

form

powder

color

orange-brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cc1cc(ccc1NC(=O)c2ccnn2C)\N=N\c3ccccc3C

InChI

1S/C19H19N5O/c1-13-6-4-5-7-17(13)23-22-15-8-9-16(14(2)12-15)21-19(25)18-10-11-20-24(18)3/h4-12H,1-3H3,(H,21,25)/b23-22+

InChI key

LKTNEXPODAWWFM-GHVJWSGMSA-N

Application

CH-223191 has been used as aryl hydrocarbon receptor (AHR) antagonist in HepaRG cells, TH17-IL-10+ cells and organoids.

Biochem/physiol Actions

CH-223191 is a potent and specific aryl hydrocarbon receptor (AhR) antagonist.
CH-223191 is a potent and specific aryl hydrocarbon receptor (AhR) antagonist. It inhibited TCDD-mediated nuclear translocation and DNA binding of AhR, and inhibited TCDD-induced luciferase activity with an IC50 of 30nM. Unlike some other AhR antagonists which display agonist activity at high concentrations, CH-223191 did not stimulate AhR-dependent transcription even at 100 micromolar. It is also specific for AhR, displaying no affinity for the estrogen receptor, as some other antagonists do.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Wang et al.
Environmental health perspectives, 121(11-12), 1334-1343 (2013-09-24)
The aryl hydrocarbon receptor (AHR) is a ligand-activated transcription factor that regulates the expression of xenobiotic detoxification genes and is a critical mediator of gene-environment interactions. Many AHR target genes identified by genome-wide gene expression profiling have morphogenetic functions, suggesting
Down-regulation of the expression of alcohol dehydrogenase 4 and CYP2E1 by the combination of alpha-endosulfan and dioxin in HepaRG human cells
Attignon E, et al.
Toxicology in vitro, 45, 309-317 (2017)
Angela Rico de Souza et al.
Frontiers in immunology, 12, 630427-630427 (2021-03-05)
Cigarette smoke is a prevalent respiratory toxicant that remains a leading cause of death worldwide. Cigarette smoke induces inflammation in the lungs and airways that contributes to the development of diseases such as lung cancer and chronic obstructive pulmonary disease
Lactobacillus accelerates ISCs regeneration to protect the integrity of intestinal mucosa through activation of STAT3 signaling pathway induced by LPLs secretion of IL-22
Hou Q, et al.
Cell Death and Differentiation, 1-1 (2018)
An immunoregulatory and tissue-residency program modulated by c-MAF in human TH 17 cells
Aschenbrenner D, et al.
Nature Immunology, 19(10), 1126-1126 (2018)

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service