C7182
Nunc® Lab-Tek® Chamber Slide™ system
8 wells, Permanox® slide, 0.8 cm2/well, sterile, 96/cs
Synonym(s):
cell culture chamber, chamber slides, microscope slides
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
Recommended Products
sterility
sterile
sterile
feature
CE marked
packaging
pack of 16
case of 96
manufacturer/tradename
Nunc 177445
well surface area
0.8 , 282
working volume
0.2-0.4 mL
suitability
suitable for (viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Nunc® Lab-Tek® Chamber Slide System
- Especially developed for microscopy of in situ tissue culture
- Cells grow on a standard microscope slide
- Growth chamber can be removed when culturing is complete
- Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
- Broad range of formats and well numbers
- Fits standard equipment
- Certified surface treatment ensures optimal contitions for cell attachment and growth
- Saves time and reagents
- Permanox® does not interfere with fluorescent labels
- No cell transfer needed prior to visualisation/staining
- Upper structure can be removed when culturing is complete
- Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
- Broad range of formats and well numbers
- Suitable for use with fluorescent labels
- CE marked
Legal Information
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Permanox is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gaston Bonenfant et al.
Journal of virology, 93(12) (2019-04-05)
Flaviviruses limit the cell stress response by preventing the formation of stress granules (SGs) and modulate viral gene expression by subverting different proteins involved in the stress granule pathway. In this study, we investigated the formation of stress granules during
Dakota J Brock et al.
Cell chemical biology, 27(10), 1296-1307 (2020-08-14)
Many cellular delivery reagents enter the cytosolic space of cells by escaping the lumen of endocytic organelles and, more specifically, late endosomes. The mechanisms involved in endosomal membrane permeation remain largely unresolved, which impedes the improvement of delivery agents. Here
Gemma Palazzolo et al.
Translational oncology, 13(4), 100760-100760 (2020-04-05)
Although screening has reduced mortality rates for colorectal cancer (CRC), about 20% of patients still carry metastases at diagnosis. Postsurgery chemotherapy is toxic and induces drug resistance. Promising alternative strategies rely on repurposing drugs such as aspirin (ASA) and metformin
Chushu Li et al.
Oncogene, 40(6), 1128-1146 (2020-12-17)
Cancer cell expression of PD-L1 leads to T cells exhaustion by transducing co-inhibitory signal, and further understanding the regulation of PD-L1 in cancer cells may provide additional therapeutic strategies. Here by drug repurposing screen, we identified amlodipine as a potent
N Joly-Tonetti et al.
The British journal of dermatology, 179(5), 1115-1126 (2018-06-30)
Melanin is synthesized by melanocytes in the basal layer of the epidermis. When transferred to surrounding keratinocytes melanin is the key ultraviolet radiation-protective biopolymer responsible for skin pigmentation. Most melanin is observable in the proliferative basal layer of the epidermis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service