Skip to Content
Merck
All Photos(1)

Key Documents

C6848

Sigma-Aldrich

Ciproxifan maleate

≥98% (HPLC), powder

Synonym(s):

Cyclopropyl (4-[3-(1H-imidazol-4-yl)propyloxy]phenyl) ketone maleate salt, FUB 359 maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2O2 · C4H4O4
CAS Number:
Molecular Weight:
386.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 32 mg/mL
H2O: insoluble

SMILES string

OC(=O)\C=C/C(O)=O.O=C(C1CC1)c2ccc(OCCCc3c[nH]cn3)cc2

InChI

1S/C16H18N2O2.C4H4O4/c19-16(12-3-4-12)13-5-7-15(8-6-13)20-9-1-2-14-10-17-11-18-14;5-3(6)1-2-4(7)8/h5-8,10-12H,1-4,9H2,(H,17,18);1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

RLQFKEYRALXXEJ-BTJKTKAUSA-N

Application

Ciproxifan maleate has been used as a selective H3R antagonist to investigate the antidepressant potential of ciproxifan in chronic unpredictable stress (CUS) model of depression in mice. It has also been used as an H3R antagonist to develop in vivo receptor occupancy assay for histamine H3 receptors (H3R)/to determine the occupancy of exogenous H3R antagonists using non-radiolabeled GSK189254 as a tracer in the rat.

Biochem/physiol Actions

Cyclopropyl 4-(3-(1H-imidazol-4-yl)propyloxy)phenyl methanone (Ciproxifan) is a species-specific histamine H3 receptor (H3R) inverse agonist. It is mainly used as a reference H3R antagonist in rodent models for neurological diseases, Alzheimer′s disease, attention deficit hyperactivity disorder (ADHD), and sleep-wake disorders. It can block monoamine oxidase A (MAO-A) and monoamine oxidase B (MAO-B) enzyme isoforms reversibly in humans and rats.
Potent, selective H3 histamine receptor antagonist.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Hagenow et al.
Scientific reports, 7, 40541-40541 (2017-01-14)
Ciproxifan is a well-investigated histamine H3 receptor (H3R) inverse agonist/antagonist, showing an exclusively high species-specific affinity at rodent compared to human H3R. It is well studied as reference compound for H3R in rodent models for neurological diseases connected with neurotransmitter
Ramakrishna Nirogi et al.
Journal of pharmacological and toxicological methods, 65(3), 115-121 (2012-04-11)
Rapid and reliable preclinical receptor occupancy measurement at the target organ in relevant species is critical in accelerating the drug hunting process. The aim of this study was to develop in vivo receptor occupancy assay for histamine H₃ receptors (H₃R)
Ajeet Kumar et al.
Psychoneuroendocrinology, 101, 128-137 (2018-11-21)
The last two decades of research has established histamine (HA) as a neurotransmitter. Since H3R antagonists are known to modulate several neurotransmitters besides HA, H3R antagonists have shown potential for the treatment of different central nervous system disorders, including depression.
X Ligneau et al.
The Journal of pharmacology and experimental therapeutics, 287(2), 658-666 (1998-11-10)
Ciproxifan, i.e., cyclopropyl-(4-(3-1H-imidazol-4-yl)propyloxy) phenyl) ketone, belongs to a novel chemical series of histamine H3-receptor antagonists. In vitro, it behaved as a competitive antagonist at the H3 autoreceptor controlling [3H]histamine release from synaptosomes and displayed similar Ki values (0.5-1.9 nM) at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service