Skip to Content
Merck
All Photos(3)

Key Documents

C6780

Sigma-Aldrich

α-Casein from bovine milk

≥70% αs-casein basis (electrophoresis), lyophilized powder

Synonym(s):

alpha-S1-casein, alpha-S2-casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

bovine milk

Quality Level

Assay

≥70% αs-casein basis (electrophoresis)

form

lyophilized powder

technique(s)

mass spectrometry (MS): suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

α-Casein from bovine milk is an Immunoglobulin E (IgE)-binding epitope belonging to the Casein phosphoprotein family. It has been used in studies of children with insulin-dependent diabetes mellitus (IDDM), due to the strong cross-reactivity of α-casein antibodies with insulin. Studies of children displaying (IDDM) show higher levels of α-casein antibodies than control siblings, which may serve as a risk marker for IDDM.

Biochem/physiol Actions

α-Casein can be an immunogen in young children, and anti-a-casein antibodies have strong cross-reactivity with insulin. This effect can explain the uncommon insulin autoimmunity, but it does not appear to progress to Type I diabetes.

Preparation Note

Chromatographically purified

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Luxenburger et al.
Molecules (Basel, Switzerland), 24(5) (2019-03-06)
In this study, we report on the modification of a 3,4-diaryl-isoxazole-based CK1 inhibitor with chiral pyrrolidine scaffolds to develop potent and selective CK1 inhibitors. The pharmacophore of the lead structure was extended towards the ribose pocket of the adenosine triphosphate
Richard D Unwin et al.
Nature protocols, 4(6), 870-877 (2009-05-16)
The application of a targeted mass spectrometric workflow to the sensitive identification of post-translational modifications is described. This protocol employs multiple reaction monitoring (MRM) to search for all putative peptides specifically modified in a target protein. Positive MRMs trigger an
K Adler et al.
Clinical and experimental immunology, 164(1), 42-49 (2011-03-03)
Insulin autoantibodies (IAA) can appear in children within months of introducing solid foods to the diet and before clinical type 1 diabetes. The aim of this study was to determine whether infant dietary antigens could be immunizing agents of IAA.
Shenmao Wu et al.
Food science and biotechnology, 28(1), 129-138 (2019-03-01)
The effect of adjunct culture Monascus purpureus BD-M-4 on the physicochemical, proteolytic, and lipolytic properties of surface mold-ripened cheese were evaluated. During the maturation of Monascus-fermented cheese, the total microbiota count and the content of soluble nitrogen increased steadily, whereas
T Saukkonen et al.
Diabetologia, 41(1), 72-78 (1998-03-14)
Dietary factors are suspected to play an aetiological role in the development of insulin-dependent diabetes mellitus (IDDM). We analysed cow's milk formula, betalactoglobulin, and bovine serum albumin antibodies by an enzyme-linked immunoassay in unselected children with newly diagnosed IDDM and

Questions

  1. 이 제픔 화학구조식과 분자량 알 수 있을까요?

    1 answer
    1. The molecular weight is approximately 23.6 kDa and will have a range of 23,000 - 26,000. There are a number of publications available on the structure, please check the literature for additional information.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service