Skip to Content
Merck
All Photos(1)

Documents

C5249

Sigma-Aldrich

Cetrorelix acetate

≥98% (HPLC)

Synonym(s):

Cetrotide, N-acetyl-3-(2-naphthalenyl)-D-alanyl-4-chloro-D-phenylalanyl-3-(3-pyridinyl)-D-alanyl-L-seryl-L-tyrosyl-N5-(aminocarbonyl)-D-ornithyl-L-leucyl-L-arginyl-L-prolyl-D-alaninamide acetic acid salt, NS-75A, SB-75

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H92ClN17O14 · xC2H4O2
CAS Number:
Molecular Weight:
1431.04 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

methanol: 10%

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](NC(=O)[C@@H](CCCNC(N)=O)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CO)NC(=O)[C@@H](Cc2cccnc2)NC(=O)[C@@H](Cc3ccc(Cl)cc3)NC(=O)[C@@H](Cc4ccc5ccccc5c4)NC(C)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N6CCC[C@H]6C(=O)N[C@H](C)C(N)=O

InChI

1S/C70H92ClN17O14.C2H4O2/c1-39(2)31-52(61(94)82-51(15-9-28-77-69(73)74)68(101)88-30-10-16-58(88)67(100)79-40(3)59(72)92)83-60(93)50(14-8-29-78-70(75)102)81-63(96)54(34-43-20-25-49(91)26-21-43)86-66(99)57(38-89)87-65(98)56(36-45-11-7-27-76-37-45)85-64(97)55(33-42-18-23-48(71)24-19-42)84-62(95)53(80-41(4)90)35-44-17-22-46-12-5-6-13-47(46)32-44;1-2(3)4/h5-7,11-13,17-27,32,37,39-40,50-58,89,91H,8-10,14-16,28-31,33-36,38H2,1-4H3,(H2,72,92)(H,79,100)(H,80,90)(H,81,96)(H,82,94)(H,83,93)(H,84,95)(H,85,97)(H,86,99)(H,87,98)(H4,73,74,77)(H3,75,78,102);1H3,(H,3,4)/t40-,50-,51+,52+,53-,54+,55-,56-,57+,58+;/m1./s1

InChI key

KFEFLCOCAHJBEA-ANRVCLKPSA-N

Gene Information

human ... GNRHR(2798)

Application

Cetrorelix, a luteinizing hormone releasing hormone receptor (LHRHR) antagonist, was used to study its safety, mechanism of action, metabolism, and efficacy as a therapeutic agent for the treatment of blood cancers such as multiple myeloma (MM). Cetrorelix is used to evaluate its effects on a wide range of physiological processes and malignancies that involve luteinizing hormone releasing hormone receptor signaling pathways involving processes such as steroidogenesis.

Biochem/physiol Actions

Cetrorelix acetate is a luteinizing hormone releasing hormone receptor antagonist. It causes prepubertal reduction in bone modeling, bone density and bone strength comparable to post-menopausal bone loss.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Laure Nivet et al.
Animal science journal = Nihon chikusan Gakkaiho, 89(1), 21-30 (2017-08-26)
In cows, the use of follicle-stimulating hormone (FSH) to stimulate follicular growth followed by a short period of FSH withdrawal has been shown to be beneficial for oocyte developmental competence. Although this treatment represents a useful optimization to generate highly
Amy Barrie et al.
Reproductive biomedicine online, 34(5), 455-462 (2017-03-21)
This retrospective, single site observational study aimed to delineate five abnormal embryonic developmental phenotypes, assessing their prevalence, development potential and suitability for inclusion in embryo selection models for IVF. In total, 15,819 embryos from 4559 treatment cycles cultured in EmbryoScope®
Bufang Xu et al.
Reproductive biology and endocrinology : RB&E, 18(1), 7-7 (2020-01-26)
Poor response patients with PCOS who are not susceptible to gonadotropin stimulation are more likely to have canceled cycles or poor clinical outcomes during IVF treatment. However, some limitations exist in the present therapies. In this study, we evaluated the
Daryosh Mohammadnejad et al.
Advanced pharmaceutical bulletin, 3(2), 323-328 (2013-12-07)
Anti cancer drugs is one of the most important chemotherapeutic factors which can influence spermatogenesis process and germinal epithelium. Since dividing cells are mainly affected by anticancer drugs, the aim of the present study is to investigate the preventive effect
José M Navarro-Pando et al.
Science advances, 7(1) (2021-02-02)
Inflammation is a hallmark of aging and is negatively affecting female fertility. In this study, we evaluate the role of the NLRP3 inflammasome in ovarian aging and female fertility. Age-dependent increased expression of NLRP3 in the ovary was observed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service