Skip to Content
Merck
All Photos(1)

Key Documents

C5151

Sigma-Aldrich

3-Carbamoyl-PROXYL

Synonym(s):

3-Carbamoyl-2,2,5,5-tetramethyl-1-pyrrolidineoxy, 3-Carbamoyl-2,2,5,5-tetramethylpyrrolidin-1-yloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17N2O2
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

173-175 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)CC(C(N)=O)C(C)(C)N1[O]

InChI

1S/C9H17N2O2/c1-8(2)5-6(7(10)12)9(3,4)11(8)13/h6H,5H2,1-4H3,(H2,10,12)

InChI key

XNNPAWRINYCIHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A spin label useful for non-invasive detection of free radicals in vivo by electron spin resonance imaging.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Th Herrling et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 63(4), 840-845 (2006-03-18)
Free radicals/reactive oxygen species (ROS) generated in skin by UV irradiation were measured by electron spin resonance (ESR). To increase the sensitivity of measurement the short life free radicals/ROS were scavenged and accumulated by using the nitroxyl probe 3-carboxy-2,2,5,5-tetrametylpyrrolidine-1-oxyl (PCA).
Ken-Ichiro Matsumoto et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(8), 2455-2462 (2006-04-28)
There is considerable research directed toward the identification and development of functional contrast agents for medical imaging that superimpose tissue biochemical/molecular information with anatomical structures. Nitroxide radicals were identified as in vivo radioprotectors. Being paramagnetic, they can provide image contrast
Guohan Hu et al.
Journal of neurosurgery, 98(2), 393-396 (2003-02-21)
Nitroxides mimic superoxide dismutase (SOD) biochemically and may prevent free radical oxidative injury in settings in which endogenous SOD is overwhelmed. The authors have previously shown the efficacy of a nitroxide, Tempol, in reducing stroke infarct size. Of the nitroxides
Tomiko Mikuni et al.
Cancer research, 64(18), 6495-6502 (2004-09-18)
Electron paramagnetic resonance imaging (EPRI) enables noninvasive spatial mapping of free radical metabolism and has recently been shown to provide in vivo physiologic information regarding alterations in the redox state of tumors and neoplastic tissues. With the use of nitroxide
T Herrling et al.
Free radical biology & medicine, 35(1), 59-67 (2003-06-27)
Reactive free radicals and reactive oxygen species (ROS) induced by ultraviolet irradiation in human skin are strongly involved in the occurrence of skin damages like aging and cancer. In the present work an ex vivo method for the detection of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service