Skip to Content
Merck
All Photos(3)

Key Documents

C4930

Sigma-Aldrich

γ-Cyclodextrin

powder, BioReagent, suitable for cell culture, ≥98%

Synonym(s):

Cyclomaltooctaose, Cyclooctaamylose, Schardinger γ-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80O40
CAS Number:
Molecular Weight:
1297.12
Beilstein:
78740
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.77

product line

BioReagent

Quality Level

Assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

1 M NaOH: 25 mg/mL, clear to slightly hazy

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@]1(O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O)O[C@@H]2[C@@H](COC(C)=O)O[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O

InChI

1S/C48H80O40/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58/h9-72H,1-8H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-/m1/s1

InChI key

GDSRMADSINPKSL-HSEONFRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclodextrins belongs to the family of cyclic α-1,4-glucans and is made of glucose units. This molecule has a shape of a hollow cone. It possesses a hydrophilic external and hydrophobic internal surface, because of which cyclodextrins can form inclusion complexes. γ-Cyclodextrin is extensively used in food and pharmaceutical industries. Cyclodextrins are obtained by the enzymatic conversion of starch/starch derivatives by cyclodextrin glycosyltransferase.

Application

A molecule used to solublize non-polar molecules such a cholesterol for use in cell culture.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

γ-Cyclodextrin: a review on enzymatic production and applications.
Li, Zhaofeng et al.
Applied Microbiology and Biotechnology, 77(2), 245-245 (2007)
Phatsawee Jansook et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 13(3), 336-350 (2010-11-26)
Study the complexation of dexamethasone in combinations of γ-cyclodextrin (γCD) and 2-hydroxypropyl-γ-cyclodextrin (HPγCD) with emphasis on solid characterization and development of aqueous dexamethasone eye drop suspension for drug delivery through sclera. Dexamethasone/cyclodextrin (dexamethasone/CD) solid complex systems were prepared and characterized
S A A Almeida et al.
Talanta, 85(3), 1508-1516 (2011-08-03)
Solid-contact sensors for the selective screening of sulfadiazine (SDZ) in aquaculture waters are reported. Sensor surfaces were made from PVC membranes doped with tetraphenylporphyrin-manganese(III) chloride, α-cyclodextrin, β-cyclodextrin, or γ-cyclodextrin ionophores that were dispersed in plasticizer. Some membranes also presented a
Sekar Karuppannan et al.
Chemistry, an Asian journal, 6(4), 964-984 (2011-01-29)
The past ten years have seen a spectacular development of chemical sensors based on the monomer-excimer dual luminescence of aromatic systems, such as pyrene. Either in the form of integrated or multicomponent molecular devices these chemosensors have been attracting a
B-G Kim et al.
British journal of pharmacology, 164(6), 1698-1710 (2011-05-19)
Although 3α-hydroxy, 5α-reduced pregnane steroids, such as allopregnanolone (AlloP) and tetrahydrodeoxycorticosterone, are endogenous positive modulators of postsynaptic GABA(A) receptors, the functional roles of endogenous neurosteroids in synaptic transmission are still largely unknown. In this study, the effect of AlloP on

Articles

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service