Skip to Content
Merck
All Photos(3)

Key Documents

C4562

Sigma-Aldrich

Monoclonal Anti-Caldesmon (Smooth) antibody produced in mouse

clone hHCD, ascites fluid

Synonym(s):

Anti-CDM, Anti-H-CAD, Anti-HCAD, Anti-L-CAD, Anti-LCAD, Anti-NAG22, Anti-h-CD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

hHCD, monoclonal

contains

15 mM sodium azide

species reactivity

sheep, rabbit, human, pig, bovine, mouse

technique(s)

immunohistochemistry: 1:500 using methacarn-fixed, paraffin-embedded sections of human or animal tissue
immunoprecipitation (IP): suitable
western blot: 1:4,000 using human uterus extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALD1(800)
mouse ... Cald1(109624)

Specificity

The antibody (also cited as h-CD) reacts in immunoblotting assays with caldesmon polypeptide of 150 kDa (h-caldesmon). It does not cross-react with skeletal or cardiac muscle or with the 70 kDa non-muscle caldesmon. In immunohistochemical staining, the antibody exhibits smooth muscle specificity. It stains vascular and visceral smooth muscle cells but not epithelial, endothelial or connective tissue fibroblast cells. In normal and malignant breast tissue, the myoepithelial component of galactophorous sinuses (but not in ducts of lobules) is stained; however, the antibody may be non-reactive by immunocytochemical methods with cultured smooth muscle cells. This probably reflects the down regulation of caldesmon commonly observed in tissue culture.

Immunogen

human uterus smooth muscle extract.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)
Monoclonal Anti-Caldesmon (Smooth) antibody produced in mouse is suitable for:
  • immunohistochemistry at a dilution of 1:500 using methacarn-fixed, paraffin-embedded sections of human or animal tissue
  • immunoprecipitation
  • western blot at a dilution of 1:4,000 using human uterus extract

Biochem/physiol Actions

Monoclonal Anti-Caldesmon (Smooth) (mouse IgG1 isotype) reacts in immunoblotting assays with caldesmon polypeptide of 150 kDa (h-Caldesmon).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa F Brereton et al.
PloS one, 8(2), e57451-e57451 (2013-02-26)
Adequate blood flow through placental chorionic plate resistance arteries (CPAs) is necessary for oxygen and nutrient transfer to the fetus and a successful pregnancy. In non-placental vascular smooth muscle cells (SMCs), K(+) channels regulate contraction, vascular tone and blood flow.
E Vardar et al.
Biomaterials, 206, 41-48 (2019-03-30)
Stress urinary incontinence (SUI) is a life changing condition, affecting 20 million women worldwide. In this study, we developed a bioactive, injectable bulking agent that consists of Permacol™ (Medtronic, Switzerland) and recombinant insulin like growth factor-1 conjugated fibrin micro-beads (fib_rIGF-1)
M G Frid et al.
Developmental biology, 153(2), 185-193 (1992-10-01)
Expression of the regulatory contractile proteins, heavy caldesmon (h-caldesmon) and calponin was studied in human aortic smooth muscle cells (SMCs) during development and compared with the expression of alpha-SM-actin and smooth muscle-myosin heavy chain (SM-MHCs). For this study, novel monoclonal
Isolation, culture, and characterization of human intestinal smooth muscle cells.
Martin Graham et al.
Methods in molecular medicine, 78, 417-423 (2003-06-27)
Nicholas S Heaton et al.
Journal of vascular research, 45(5), 365-374 (2008-03-21)
The success of peripheral vein grafts is limited by intimal hyperplasia. Transforming growth factor (TGF)-beta(1) has effects on cell proliferation, apoptosis and extracellular matrix synthesis. We have previously observed positive changes in vessel healing with antisense to TGF-beta(1). Adenovirus was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service