C4418
L-Cysteinesulfinic acid monohydrate
Synonym(s):
3-Sulfinoalanine
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Empirical Formula (Hill Notation):
C3H7NO4S · H2O
CAS Number:
Molecular Weight:
171.17
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
form
powder
Quality Level
storage temp.
−20°C
SMILES string
NC(CS(O)=O)C(O)=O
InChI
1S/C3H7NO4S/c4-2(3(5)6)1-9(7)8/h2H,1,4H2,(H,5,6)(H,7,8)
InChI key
ADVPTQAUNPRNPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
L-Cysteinesulfinic acid monohydrate has been used in the fluorescence measurement of cysteine oxidation.
Biochem/physiol Actions
Cysteinesulfinic acid (CSA), also called 3-sulfinoalanine is synthesized by the conversion of sulfur to sulfinic acid in cysteine by the enzyme cysteine dioxygenase (CDO). CSA is a putative excitatory amino acid (EAA) with neurotransmitter functionality. It also regulates cardiovascular functions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cardiovascular actions of L-cysteine and L-cysteine sulfinic acid in the nucleus tractus solitarius of the rat
Takemoto Y
Amino Acids, 46(7), 1707-1713 (2014)
Oxidation of hypotaurine and cysteine sulphinic acid by peroxynitrite
Fontana M, et al.
The Biochemical Journal, 389(1), 233-240 (2005)
Cysteine sulphinate (CSA) as an excitatory amino acid transmitter candidate in the mammalian central nervous system.
R Griffiths
Progress in neurobiology, 35(4), 313-323 (1990-01-01)
Cysteine dioxygenase: a robust system for regulation of cellular cysteine levels
Stipanuk MH, et al.
Amino Acids, 37(1), 55-55 (2009)
N-acetyl cysteine functions as a fast-acting antioxidant by triggering intracellular H2S and sulfane sulfur production
Ezericna D, et al.
Cell Chemical Biology, 25(4), 447-459 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service