Skip to Content
Merck
All Photos(1)

Documents

C3743

Sigma-Aldrich

CI 976

>98% (HPLC), solid

Synonym(s):

2,2-Dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H39NO4
CAS Number:
Molecular Weight:
393.56
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: insoluble <2 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC(C)(C)C(=O)Nc1c(OC)cc(OC)cc1OC

InChI

1S/C23H39NO4/c1-7-8-9-10-11-12-13-14-15-23(2,3)22(25)24-21-19(27-5)16-18(26-4)17-20(21)28-6/h16-17H,7-15H2,1-6H3,(H,24,25)

InChI key

WAFNZAURAWBNDZ-UHFFFAOYSA-N

Application

CI 976 has been used as an acyl-coenzyme A: cholesterol acyltransferase (ACAT) inhibitor:
  • to analyze its anti-hepatitis C virus (HCV) activity in Huh7.5.1 cells
  • to treat Neuro-2a cells to test its effect on plasma membrane integrated density of α4-SEPβ2 or α6-SEPβ2β3 nicotinic acetylcholine receptors (nAChRs)
  • to study its effects on the anti-angiogenic activity of pyripyropenes in human umbilical vein endothelial cells

Biochem/physiol Actions

CI-976, a new trimethoxy fatty acid anilide, is a potent and specific inhibitor of liver and intestinal acyl coenzyme A; cholesterol acyltransferase (ACAT) in vitro. CI-976 decreased non-high density lipoprotein (HDL)-cholesterol and increased HDL-cholesterol in rats with pre-established dyslipidemia. High performance gel chromatographic separation of plasma lipoproteins also revealed that CI-976, but not CL 277,082, lowered low density lipoprotein (LDL)-cholesterol and elevated HDL-cholesterol.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asami Hayashi et al.
Biological & pharmaceutical bulletin, 32(7), 1261-1265 (2009-07-03)
In the course of our search for anti-angiogenic substances, pyripyropenes A (1), B (2), and D (3) were re-discovered as selective anti-proliferative substances against human umbilical vein endothelial cells (HUVECs) from a marine-derived fungus of Aspergillus sp. Pyripyropenes showed potent
S Sonda et al.
The Journal of biological chemistry, 276(37), 34434-34440 (2001-07-18)
Upon host cell invasion the apicomplexan parasite Toxoplasma gondii resides in a specialized compartment termed the parasitophorous vacuole that is derived from the host cell membrane but modified by the parasite. Despite the segregation of the parasitophorous vacuole from the
Daniel Drecktrah et al.
Molecular biology of the cell, 14(8), 3459-3469 (2003-08-20)
Recent studies have suggested that formation of Golgi membrane tubules involves the generation of membrane-associated lysophospholipids by a cytoplasmic Ca2+-independent phospholipase A2 (PLA2). Herein, we provide additional support for this idea by showing that inhibition of lysophospholipid reacylation by a
R A Harte et al.
Biochimica et biophysica acta, 1258(3), 241-250 (1995-10-05)
The effect of the membrane environment of acyl-CoA:cholesterol acyl transferase (ACAT), an important intracellular enzyme of cholesterol metabolism, on the properties of a range of inhibitors of varying potencies was studied. ACAT activity from rat liver was solubilised with 3%
Y Sakuma et al.
Japanese journal of pharmacology, 70(1), 35-41 (1996-01-01)
FR129169 (FR) (N-(1,2-diphenylethyl)-2-octyloxyphenylacetamide) has been found to inhibit acyl-CoA:cholesterol acyltransferase (ACAT) activities in intestinal microsomes of rats and rabbits and the liver homogenate of rats with IC50 values of around 1.0 x 10(-7) M. The inhibitory activity was 2-3 times

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service