Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
C3175
Chloramphenicol-Water Soluble
powder, suitable for cell culture, BioReagent
Synonym(s):
Chloramphenicol, Chlornitromycin, Chlorocid, Chloromycetin, Cloramfenicol, D-(−)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-(−)-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, Chloromycetin, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]aceta, Kloramfenikol
About This Item
Recommended Products
Product Name
Chloramphenicol-Water Soluble, powder, BioReagent, suitable for cell culture
product line
BioReagent
form
powder
technique(s)
cell culture | mammalian: suitable
solubility
H2O: 50-500 mg/mL (stock solution)
PBS: 50-500 mg/mL (stock solution)
other salt solutions: not recommended
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma
Mode of action
protein synthesis | interferes
Application
Biochem/physiol Actions
Packaging
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic
Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic
Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic
Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
How do I calculate the molarity of product C3175, Chloramphenicol-Water Soluble?
1 answer-
Product C3175 (Chloramphenicol-Water Soluble) is a complex of chloramphenicol and cyclodextrin. Cyclodextrin is used to aid in the solubility of chloramphenicol into water. In order to prepare a molar solution of chloramphenicol you will have to base it on the chloramphenicol content in the solution and not the entire complex. Product C3175 is formulated to contain approximately 100 mg chloramphenicol per gram of powder. Based on this average, if you are preparing a 500 mg/ml stock solution, there will be approximately 50 mg of chloramphenicol in 1 ml. Below is the calculated molarity of chloramphenicol when the complex is dissolved at 500 mg/ml in water. molecular weight X unknown molarity X volume = complex weight X complex ratio323.13 g/mol X Molarity X 1 ml = 500 mg X 0.1 Molarity = 0.155 Mchloramphenicol molecular weight = 323.13 g/molmolarity (unknown) = mol/Lvolume = 1mlcomplex weight = 500 mgcomplex ratio = 100 mg chloramphenicol/1 gm complex
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service