Skip to Content
Merck
All Photos(1)

Key Documents

C3117

Sigma-Aldrich

Monoclonal Anti-Cy3/Cy5−Biotin antibody produced in mouse

~1 mg/mL, clone CY-96, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-Cy3/Cy5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CY-96, monoclonal

form

buffered aqueous solution

concentration

~1 mg/mL

technique(s)

direct ELISA: suitable
dot blot: suitable
immunocytochemistry: 1-2 μg/mL using chicken fibroblasts
microarray: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Cy3/Cy5 (mouse IgG2a isotype), Biotin conjugate is derived from the CY-96 hybridoma produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with a mixture of proteins labelled with cyanine 3 (Cy3) or Cy5. Cyanine dyes are ionized polymethine dyes.

Specificity

The antibody recognizes Cy3 and Cy5 conjugated to proteins.

Immunogen

mixture of proteins labeled with Cy3/Cy5.

Application

Monoclonal Anti-Cy3/Cy5 Biotin antibody produced in mouse has also been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • dot blot assay
  • protein microarrays
  • immunofluorescenc

Monoclonal Anti-Cy3/Cy5-Biotin antibody produced in mouse was used to label miR-107 in human breast cancer cell lines by confocal microscopy.

Biochem/physiol Actions

Cy dyes are pH insensitive fluorophores that are used to label, DNA, RNA and proteins in a variety of biological assays.
Cyanine dyes are small, soluble in aqueous solution and has tolerance to dimethyl sulfoxide (DMSO). They are more photostable than fluorescein, has high molar extinction coefficients and favourable quantum yields. Monoclonal antibodies to cyanine 3 (Cy3) and/or Cy5 are an important tool for Cy3/Cy5 signal amplification in assays where biomolecules are labelled with Cy3 or Cy5.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Animal models and experimental medicine (2023-08-11)
The effect of platelet factor 4 (PF4) on bone marrow mesenchymal stem cells (BMMSCs) and osteoporosis is poorly understood. Therefore, this study aimed to evaluate the effects of PF4-triggered bone destruction in mice and determine the underlying mechanism. First, in
Quantitative profile of the uropathogenic Escherichia coli outer membrane proteome during growth in human urine
Alteri CJ and Mobley HLT
Infection and Immunity, 75(6), 2679-2688 (2007)
Chao-Yue Yang et al.
Chinese journal of traumatology = Zhonghua chuang shang za zhi, 26(3), 155-161 (2023-04-06)
This study aims to elucidate the electrotaxis response of alveolar epithelial cells (AECs) in direct-current electric fields (EFs), explore the impact of EFs on the cell fate of AECs, and lay the foundation for future exploitation of EFs for the
Yejun Tan et al.
Frontiers in immunology, 14, 1149810-1149810 (2023-04-11)
Patients with diabetes mellitus (DM) have a higher incidence of malignant tumors than people without diabetes, but the underlying molecular mechanisms are still unclear. To investigate the link between DM and cancer, we screened publicly available databases for diabetes and
25th Anniversary Article: Design of Polymethine Dyes for All-Optical Switching Applications: Guidance from Theoretical and Computational Studies
Gieseking RL, et al.
Advanced Materials, 26(1), 68-84 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service