Skip to Content
Merck
All Photos(1)

Key Documents

C191

Sigma-Aldrich

Capsazepine

≥98% (HPLC), solid, TRPV1 antagonist

Synonym(s):

N-[2-(4-Chlorophenyl)ethyl]-1,3,4,5-tetrahydro-7,8-dihydroxy-2H-2-benzazepine-2-carbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21ClN2O2S
CAS Number:
Molecular Weight:
376.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Capsazepine, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Oc1cc2CCCN(Cc2cc1O)C(=S)NCCc3ccc(Cl)cc3

InChI

1S/C19H21ClN2O2S/c20-16-5-3-13(4-6-16)7-8-21-19(25)22-9-1-2-14-10-17(23)18(24)11-15(14)12-22/h3-6,10-11,23-24H,1-2,7-9,12H2,(H,21,25)

InChI key

DRCMAZOSEIMCHM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Capsazepine has been used as a transient receptor potential vanilloid-1 (TRPV1) antagonist:
  • to study its effect on capsaicin induced extracellular signal-regulated kinase (ERK) phosphorylation[1]
  • to study the role of TRPV1 in central terminals on nociception in rats[2]
  • for functional characterization of the TRPV1 in bull spermatozoa[3]

Biochem/physiol Actions

Capsazepine is a synthetic analog of capsaicin and a transient receptor potential vanilloid-1 (TRPV1) antagonist.[1] It exhibits anti-proliferative and anti-cancer effects in various cancer types including oral squamous cell carcinoma, non-small cell lung cancer (NSCLC), breast cancer, and prostate cancer cell lines (HSC-3, H460, MDA-231, and PC-3 respectively).[4] Capsazepine recovers impaired lung mechanics during endotoxemia and it may be a potential therapeutic target for acute lung injury (ALI).[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugo Balleza-Tapia et al.
eLife, 7 (2018-11-13)
Amyloid-β peptide (Aβ) forms plaques in Alzheimer's disease (AD) and is responsible for early cognitive deficits in AD patients. Advancing cognitive decline is accompanied by progressive impairment of cognition-relevant EEG patterns such as gamma oscillations. The endocannabinoid anandamide, a TrpV1-receptor
Ablation and regeneration of peripheral and central TRPV1 expressing nerve terminals and the consequence of nociception
Yu SQ and Premkumar LS
Open Medicine : A Peer-Reviewed, Independent, Open-Access Journal, 8(1) (2015)
S Bevan et al.
British journal of pharmacology, 107(2), 544-552 (1992-10-01)
1. Capsazepine is a synthetic analogue of the sensory neurone excitotoxin, capsaicin. The present study shows the capsazepine acts as a competitive antagonist of capsaicin. 2. Capsazepine (10 microM) reversibly reduced or abolished the current response to capsaicin (500 nM)
Jorge De La Chapa et al.
Bioorganic & medicinal chemistry, 27(1), 208-215 (2018-12-12)
We previously demonstrated that capsazepine (CPZ), a synthetic transient receptor potential Vanilloid subtype 1 (TRPV1) antagonist, has significant anti-cancer effects in vivo. The purpose of this study was to develop more potent analogs based upon CPZ pharmacophore and structure-activity relationships
Layla D M Cabral et al.
Basic & clinical pharmacology & toxicology, 119(5), 421-427 (2016-04-20)
Acute lung injury (ALI) caused by systemic inflammatory response remains a leading cause of morbidity and mortality in critically ill patients. Management of patients with sepsis is largely limited to supportive therapies, reflecting an incomplete understanding of the underlying pathophysiology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service