Skip to Content
Merck
All Photos(2)

Key Documents

C1688

Sigma-Aldrich

Monoclonal Anti-C-Reactive Protein antibody produced in mouse

clone CRP-8, ascites fluid

Synonym(s):

Anti-CRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

CRP-8, monoclonal

mol wt

antigen 24 kDa

contains

15 mM sodium azide

species reactivity

human

technique(s)

dot blot: suitable
indirect ELISA: 1:40,000
western blot: 1:200

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRP(1401)

Related Categories

General description

C-reactive protein (CRP) is a member of the pentraxin family of proteins and synthesized by the liver. It is also known as PTX1. The CRP gene is mapped to human chromosome 1q23.2.

Specificity

The antibody recognizes an epitope located on the 24 kDa subunit of denatured and reduced CRP. It does not cross-react with human serum amyloid P component (SAP), human haptoglobin, human α1-acid glycoprotein, human IgG, and CRP from Limulus.

Immunogen

human C-reactive protein.

Application

  • It has been used in immunoblotting assay, to identify monomeric CRP.
  • In time-resolved fluorometric assay.
Monoclonal Anti-C-Reactive Protein antibody produced in mouse is suitable for the following:
  • dot blot analysis
  • indirect ELISA at a working dilution of 1:40,000
  • western blotting at a working dilution of 1:200

Biochem/physiol Actions

C-reactive protein (CRP) shows essential scavenging and metabolic activity and is also associated with defence mechanism. It activates the complement pathway by binding to calcium-dependent exogenous and autologous molecules containing phosphocholine (PC). It may have pathogenic effects and is a practical prognostic marker in patients with hepatocellular carcinoma (HCC). The CRP diagnostic value is higher for acute pyelonephritis than lower urinary tract infection. Thus, it helps in differentiating between upper and lower urinary tract infection. It is considered best for predicting the risk of cardiovascular disease..

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgit Fendl et al.
Scientific reports, 11(1), 6996-6996 (2021-03-28)
There is increasing evidence that C-reactive protein (CRP) can mediate inflammatory reactions following the transformation of functionally inert pentameric CRP (pCRP) into its structural isoform pCRP* and into monomeric CRP (mCRP). This conversion can occur on the membranes of apoptotic
Se Jin Oh et al.
Interactive cardiovascular and thoracic surgery, 25(2), 260-267 (2017-05-06)
We evaluated the effect of monomeric C-reactive protein (CRP) deposition on areas at risk (AAR) of myocardium with ischaemia-reperfusion injury. Myocardial ischaemia-reperfusion injury model was produced by ligation of the left anterior descending coronary artery for 45 min followed by 45 min
C-reactive protein specifically enhances platelet-activating factor-induced inflammatory activity in vivo.
Sato A, et al.
European Journal of Pharmacology, 745, 46-51 (2014)
Eun Na Kim et al.
Journal of chest surgery, 54(1), 9-16 (2021-03-27)
The deposition of monomeric C-reactive protein (mCRP) in the myocardium aggravates ischemia-reperfusion injury (IRI) and myocardial infarction. Ischemic preconditioning (IPC) is known to protect the myocardium against IRI. We evaluated the effects of IPC on myocardium upon which mCRP had
Lin Zhang et al.
Frontiers in immunology, 9, 511-511 (2018-03-30)
Native C-reactive protein (nCRP) is a non-specific marker of inflammation being claimed as a bystander in several chronic disorders. Accumulating evidence indicates that nCRP dissociates to and acts primarily as the monomeric conformation (mCRP) at inflammatory loci. This suggests that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service