Skip to Content
Merck
All Photos(2)

Key Documents

C0609

Sigma-Aldrich

Carnosic acid from Rosmarinus officinalis

≥91%, powder

Synonym(s):

(4AR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥91%

form

powder

color

faintly yellow to dark yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)c1cc2CC[C@H]3C(C)(C)CCC[C@]3(C(O)=O)c2c(O)c1O

InChI

1S/C20H28O4/c1-11(2)13-10-12-6-7-14-19(3,4)8-5-9-20(14,18(23)24)15(12)17(22)16(13)21/h10-11,14,21-22H,5-9H2,1-4H3,(H,23,24)/t14-,20+/m0/s1

InChI key

QRYRORQUOLYVBU-VBKZILBWSA-N

Related Categories

Application

Carnosic acid from Rosmarinus officinalis is used to study its mechanisms as an antioxidant that protects lipids from peroxidation and proteins from carbonylation. Carnosic acid is used to assess its neuroprotective activies including its ability to modulate the neurotoxic effects of 6-hydroxydopamine (6-OHDA). Carnosic acid may also be used to study it anticancer potential.
Phenolic compound that is naturally occurring with antioxidant properties. Inhibits lipid peroxidation which is caused by NADH or NADPH oxidation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Saxe Einbond et al.
Fitoterapia, 83(7), 1160-1168 (2012-07-26)
Studies indicate that extracts and purified components, including carnosic acid, from the herb rosemary display significant growth inhibitory activity on a variety of cancers. This paper examines the ability of rosemary/carnosic acid to inhibit the growth of human breast cancer
Tayebeh Rezaie et al.
Investigative ophthalmology & visual science, 53(12), 7847-7854 (2012-10-20)
The herb rosemary has been reported to have antioxidant and anti-inflammatory activity. We have previously shown that carnosic acid (CA), present in rosemary extract, crosses the blood-brain barrier to exert neuroprotective effects by upregulating endogenous antioxidant enzymes via the Nrf2
Mariano Walter Pertino et al.
Journal of natural products, 73(4), 639-643 (2010-04-03)
Carnosic acid (1) has been shown to possess gastroprotective activity in vitro and in vivo. However, little is known of the gastroprotective effect or cytotoxicity of carnosic acid gamma-lactone (3). To determine structure-activity relationships, a series of 17 esters of
Riham Salah El Dine et al.
Journal of natural products, 72(11), 2019-2023 (2009-10-10)
A new farnesyl hydroquinone, ganomycin I (1), was isolated along with ganomycin B (2) from the chloroform extract of the fruiting bodies of the Vietnamese mushroom Ganoderma colossum. These compounds inhibited HIV-1 protease with IC50 values of 7.5 and 1.0
A I Kuzmenko et al.
Journal of photochemistry and photobiology. B, Biology, 48(1), 63-67 (1999-04-17)
Lipid free-radical oxidation has been studied in vivo in the mitochondrial fractions of the liver of rats fed RoseOx (carnosic acid nutritional supplement) by measuring chemiluminescence. The kinetics of the lipid chemiluminescence in rats fed RoseOx are significantly different from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service