Skip to Content
Merck
All Photos(2)

Documents

B8894

Sigma-Aldrich

Albumin, Acetylated from bovine serum

protease free, for molecular biology

Synonym(s):

Ac-BSA, Acetyl BSA, Acetylated BSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

grade

for molecular biology

product line

BioReagent

form

solution

purified by

heat shock fractionation

contains

does not contain stabilizer or preservative

origin

USA origin

concentration

20 mg/mL in H2O

technique(s)

FISH: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

UniProt accession no.

foreign activity

DNase free
DNase, RNase, NICKase and protease, none detected
NICKase free
Protease free
RNase free

storage temp.

−20°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Serum albumin is the most extensive plasma protein in the circulatory system. It is produced within the liver and exported as a non-glycosylated protein to the blood plasma. The molecular weight of native serum albumin is around 66 kDa. Native serum albumin is a negatively charged protein.

Acetylation of bovine serum albumin particularly modifies lysine (Lys) residues with elevated frequency, with lesser degrees of modification of serine (Ser) and threonine (Thr) residues. MALDI-TOF mass spectrometry analysis of acetylated BSA has indicated an increased MW of about 3000, compared to native BSA (Ostatná, V. et al., J. Electroanalytical Chem., 821, 97-103 (2018)).. This acetylation process also derivatizes similar amino acid residues in any trace contaminating nucleases in the BSA, to inactivate that nuclease activity. This makes acetylated BSA useful in molecular biology applications where trace nuclease activity must be minimized.

Application

Acetylated BSA has been used as a component of the reaction mixture for adapter ligation. In addition, this B8894 acetylated BSA product has been used in other applications, such as:
  • In cDNA library preparation (Hafner, M. et al., RNA, 17(9), 1697-1712 (2011))
  • As a blocking agent with streptavidin beads used for capture of biotinylated RNA (Winz, M.-L. et al., Nat. Protocol., 12(1), 122-149 (2017))
Acetylated to inactivate nucleases commonly found in BSA. Acetylated BSA should not be used as a protein standard since acetylation of tyrosine residues prevents color development in the Lowry assay and other similar protein determinations.

Biochem/physiol Actions

Native serum albumin plays a major role in maintaining the pH and osmotic pressure of the blood. It is also involved in transporting a diverse range of endogenous and exogenous metabolites, such as amino acids, fatty acids, drugs, steroids and metal ions. Native serum albumin can have a role in modulating the levels of Ca2+ and Mg2+ metals in blood.

Features and Benefits

This acetylated BSA product has been tested for the following properties:
  • DNase-free / Exonuclease-free
  • Nickase-free / Endonuclease-free
  • Protease-free
  • RNase-free

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan Haralampiev et al.
Nature communications, 11(1), 4355-4355 (2020-08-30)
The genome of influenza A viruses (IAV) is encoded in eight distinct viral ribonucleoproteins (vRNPs) that consist of negative sense viral RNA (vRNA) covered by the IAV nucleoprotein. Previous studies strongly support a selective packaging model by which vRNP segments are
Hongshan Guo et al.
Nature protocols, 10(5), 645-659 (2015-04-04)
The heterogeneity of DNA methylation within a population of cells necessitates DNA methylome profiling at single-cell resolution. Recently, we developed a single-cell reduced-representation bisulfite sequencing (scRRBS) technique in which we modified the original RRBS method by integrating all the experimental
Xiang Yu et al.
STAR protocols, 2(4), 100901-100901 (2021-11-25)
Eukaryotic RNAs can be modified with a non-canonical 5' nicotinamide adenine dinucleotide (NAD+) cap. NAD-seq identifies transcriptome-wide NAD+ capped RNAs. NAD-seq takes advantage of click chemistry to allow the capture of NAD+ capped RNAs. Unlike other approaches, NAD-seq does not
Catherine Chailleux et al.
Nature protocols, 9(3), 517-528 (2014-02-08)
Recent advances in our understanding of the management and repair of DNA double-strand breaks (DSBs) rely on the study of targeted DSBs that have been induced in living cells by the controlled activity of site-specific endonucleases, usually recombinant restriction enzymes.
Anna Maria Sole Giordano et al.
The Journal of experimental medicine, 219(4) (2022-03-10)
Aberrant induction of type I IFN is a hallmark of the inherited encephalopathy Aicardi-Goutières syndrome (AGS), but the mechanisms triggering disease in the human central nervous system (CNS) remain elusive. Here, we generated human models of AGS using genetically modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service