Skip to Content
Merck
All Photos(1)

Key Documents

B6901

Sigma-Aldrich

Monoclonal Anti-Bovine IgG antibody produced in mouse

clone BG-18, ascites fluid

Synonym(s):

Monoclonal Anti-Bovine IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

BG-18, monoclonal

contains

15 mM sodium azide

species reactivity

bovine (non-reactive to IgM)

should not react with

pig, human, rabbit, sheep, cat, guinea pig, canine, horse, goat

technique(s)

dot blot: suitable
indirect ELISA: 1:1000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The antibody reacts with an epitope on both bovine IgG1 and IgG2 in their native form. The antibody shows no reaction with the epitope after denaturation and reduction.
The antibody reacts with an epitope on both bovine IgG1 and IgG2 in their native form. The antibody shows no reaction with the epitope after denaturation and reduction. IgG (immunoglobulin G) contributes to 10–20% of plasma protein and is regarded as one of the most predominant serum protein. It consists of four subclasses: IgG1, IgG2, IgG3 and IgG4. The IgG structure possesses four polypeptide chains containing two identical γ heavy (H) chains and two identical γ or γ light (L) chains of 50 kDa and 25 kDa, respectively.

Specificity

bovine IgG1 and IgG2

Immunogen

Purified bovine IgG

Application

Monoclonal Anti-Bovine IgG antibody has been used in indirect immunoperoxidase staining.
Monoclonal Anti-Bovine IgG antibody produced in mouse is suitable for ELISA at a working dilution of 1:20,000. It may be used for immunohistochemistry of formalin-fixed, paraffin-embedded sections at a dilution of 1:1000. Monoclonal Anti-Bovine IgG antibody produced in mouse was used as an isotype control in ChIP assay.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Ascites fluid containing 0.1% sodium azide

Storage and Stability

For continuous use, store at 2-8°C. For extended storage, solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chika Okafor et al.
Sensors (Basel, Switzerland), 8(9), 6015-6025 (2008-09-26)
Johne's disease (JD) is one of the most costly bacterial diseases in cattle. In the U.S., economic losses from the disease have been estimated to exceed $1,500,000,000 per year, mainly from the effects of reduced milk production. Current diagnostic tests
Clare F J Grant et al.
Journal of virology, 91(9) (2017-02-24)
Foot-and-mouth disease virus (FMDV) is a highly contagious viral disease. Antibodies are pivotal in providing protection against FMDV infection. Serological protection against one FMDV serotype does not confer interserotype protection. However, some historical data have shown that interserotype protection can
B Gottstein et al.
International journal for parasitology, 28(4), 679-691 (1998-05-29)
Neospora caninum has gained considerable attention through its role in the aetiology of bovine abortion. Due to its close phylogenetic relationship with Toxoplasma gondii, respective unequivocal differential diagnosis deserves special consideration. In order to evaluate the diagnostic performance of molecular
Histological and Immunohistochemical Studies on the Furstenberg's Rosette in Cows.
Asti R N, et al.
Kafkas U?niversitesi veteriner faku?ltesi dergisi, 17(2), 520-520 (2011)
Mojtaba Porbahaie et al.
Nutrients, 14(21) (2022-11-12)
Bovine milk IgG (bIgG) was shown to bind to and neutralize the human respiratory synovial virus (RSV). In animal models, adding bIgG prevented experimental RSV infection and increased the number of activated T cells. This enhanced activation of RSV-specific T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service