Skip to Content
Merck
All Photos(1)

Key Documents

B5926

Sigma-Aldrich

10× REDTaq® PCR Reaction Buffer

To be used with REDTaq® DNA Polymerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106306
NACRES:
NA.52

Quality Level

form

liquid

color

colorless

storage temp.

−20°C

General description

10x REDTaq® PCR Reaction Buffer is a polymerase chain reaction buffer for use with REDTaq® DNA Polymerase (D4309).

Application

10× REDTaq® PCR Reaction Buffer has been used as a component of reaction mix:

  • for PCR detection of virulence genes from Campylobacter jejuni food and clinical isolates in BALB/c mice
  • for PCR detection of virulence genes of Campylobacter coli isolates from infected mice liver
  • for identifying K-ras mutated alleles by PCR-restriction fragment length polymorphism (RFLP) in patients with colorectal carcinoma

Legal Information

REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin M Symonds et al.
Applied and environmental microbiology, 75(5), 1402-1409 (2009-01-07)
Human fecal matter contains a large number of viruses, and current bacterial indicators used for monitoring water quality do not correlate with the presence of pathogenic viruses. Adenoviruses and enteroviruses have often been used to identify fecal pollution in the
Romain Marti et al.
Applied and environmental microbiology, 75(15), 4967-4974 (2009-06-16)
The objective of this study was to identify a microbial marker for pig manure contamination. We quantified the persistence of four dominant bacterial groups from the pig intestinal tract throughout manure handling at 10 livestock operations (including aerobic digestion) by
Chris Waskewich et al.
Cancer research, 62(7), 2029-2033 (2002-04-04)
Cyclooxygenase-2 (COX-2) is an important cellular target for both therapy and/or prevention of inflammatory disorders and cancer. The advent of selective COX-2 inhibitors now allows a more precise and safer treatment approach. The screening of an array of cancer cell
Ovidiu Paun et al.
Molecular biology and evolution, 27(11), 2465-2473 (2010-06-17)
Epigenetic information includes heritable signals that modulate gene expression but are not encoded in the primary nucleotide sequence. We have studied natural epigenetic variation in three allotetraploid sibling orchid species (Dactylorhiza majalis s.str, D. traunsteineri s.l., and D. ebudensis) that
Christoph P Dieterle et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 10(2), 641-650 (2004-02-05)
The aim of this study was to identify K-ras mutations as marker for isolated tumor cells in liver, lymph node, and bone marrow specimens of colorectal cancer patients. To detect these, a PCR-RFLP assay was used with a sensitivity exceeding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service