Skip to Content
Merck
All Photos(2)

Documents

B4014

Sigma-Aldrich

Brilliant Green

Dye content, ≥90%, certified by the Biological Stain Commission, powder

Synonym(s):

Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
Colour Index Number:
42040
Beilstein:
3901207
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Brilliant Green, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

form

powder

composition

Dye content, ≥90%

color

dark green

mp

210 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL, blue

εmax

≥1750 at 628- 632  nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OS([O-])(=O)=O.CCN(CC)c1ccc(cc1)C(\c2ccccc2)=C3/C=CC(\C=C3)=[N+](/CC)CC

InChI

1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

NNBFNNNWANBMTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for bacteriostatic potency. Tested as a bacteriostatic agent against a spore-former and two members of the colon-typhoid group.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosef D Huberman et al.
Poultry science, 98(6), 2422-2431 (2019-01-29)
Salmonellosis caused by Salmonella Enteritidis is a widespread zoonosis and poultry products are an important source of infection. This study was carried out to evaluate the protection of different vaccination schedules in layers using a live commercial attenuated Salmonella Enteritidis
Wendy C Andersen et al.
Analytica chimica acta, 637(1-2), 279-289 (2009-03-17)
Liquid chromatographic methods are presented for the quantitative and confirmatory determination of crystal violet (CV; also known as gentian violet), leucocrystal violet (LCV), brilliant green (BG), and leucobrilliant green (LBG) in catfish. LCV and LBG were oxidized to the chromic
Mohammad Saeid Hosseini et al.
Journal of hazardous materials, 169(1-3), 52-57 (2009-04-22)
A new chelating polymeric sorbent as an extractant-impregnated resin (EIR) has been developed using brilliant green (BG) and Amberlite XAD-7 resin. The BG-impregnated resin showed superior binding affinity for Cr(VI) in the presence of many co-existing ions and no considerable
Efficient fluorescence enhancement and cooperative binding of an organic dye in a supra-biomolecular host-protein assembly.
Achikanath C Bhasikuttan et al.
Angewandte Chemie (International ed. in English), 46(22), 4120-4122 (2007-04-20)
Zarrin Es'haghi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(3), 603-607 (2011-05-03)
This paper describes the development of a new design of hollow fiber solid/liquid phase microextraction (HF-SLPME) for determination of brilliant green (BG) residues in water fish ponds. This method consists of an aqueous donor phase and carbon nanotube reinforced organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service