Skip to Content
Merck
All Photos(1)

Documents

B3759

Sigma-Aldrich

Anti-Bovine Serum antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti-Serum, bovine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

bovine

packaging

vial of 2 mL lyophilized antiserum

technique(s)

immunoelectrophoresis: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)

Looking for similar products? Visit Product Comparison Guide

Application

Anti-Bovine Serum antibody produced in rabbit has been used in differential staining.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Storage and Stability

Prior to reconstitution store the product at 2-8 °C. After reconstitution, the solution may be stored frozen in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachariah McLean et al.
Biology of reproduction, 91(2), 49-49 (2014-07-11)
Mammalian blastocysts comprise three distinct lineages, namely, trophoblast, hypoblast, and epiblast, which develop into fetal placenta, extraembryonic yolk sac, and embryo proper, respectively. Pluripotent embryonic stem cells, capable of forming all adult cell types, can only be derived from the
Increased MAP kinase inhibition enhances epiblast-specific gene expression in bovine blastocysts
McLean Z, et al.
Biology of Reproduction, 91(2), 49-41 (2014)
Efficient production of GGTA1 knockout porcine embryos using a modified handmade cloning (HMC) method
Kumbha R, et al.
Research in Veterinary Science, 128, 59-68 (2020)
Dongxia Hou et al.
PloS one, 10(11), e0142442-e0142442 (2015-11-12)
Trophoblasts (TR) are specialized cells of the placenta and play an important role in embryo implantation. The in vitro culture of trophoblasts provided an important tool to investigate the mechanisms of implantation. In the present study, porcine trophoblast cells were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service